Skip to Content
Merck
All Photos(2)

Key Documents

HPA028699

Sigma-Aldrich

Anti-FLG2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-IFPS, Anti-filaggrin family member 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Estimated to ship on14 April 2025



Select a Size

Change View
100 μL
$952.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$952.00


Estimated to ship on14 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200-1:500

immunogen sequence

CGYSNSSGCGRPQNASSSCQSHRFGGQGNQFSYIQSGCQSGIKGGQGHGCVSGGQPSGCGQPESNPCSQSYSQRGYGARENGQPQNCGGQWRTGSSQSSCCG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FLG2(388698)

General description

Filaggrin-2 (FLG2) is a 248 kDa S100 fused-type protein, that is insoluble in water. It is produced by granular keratinocytes and has 23 homologous repeats and a S100-homologous N-terminal domain. It is located in keratohyalin granules and the cytoplasmic matrix of the lower corneocytes. FLG2 is located on human chromosome 1q21.3.

Immunogen

filaggrin family member 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Filaggrin-2 (FLG2) may be involved in epithelial cornification. It plays an important role in the epidermal barrier functions. Mutation in FLG2 gene is associated with peeling skin syndrome.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76869

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deimination of human filaggrin-2 promotes its proteolysis by calpain 1
Hsu CY, et al.
The Journal of Biological Chemistry, jbc-M110 (2011)
Peeling skin syndrome associated with novel variant in FLG2 gene.
Alfares A, et al.
American Journal of Medical Genetics. Part A, 173(12), 3201-3204 (2017)
Skin-Derived C-Terminal Filaggrin-2 Fragments Are Pseudomonas aeruginosa-Directed Antimicrobials Targeting Bacterial Replication.
Hansmann B, et al.
PLoS Pathogens, 11(9), e1005159-e1005159 (2015)
Daphne P C Vergouwen et al.
Investigative ophthalmology & visual science, 64(3), 27-27 (2023-03-18)
Scleritis is a severe inflammatory ocular disorder with unknown pathogenesis. We investigated healthy sclera as well as sclera affected by noninfectious scleritis for differentially expressed proteins using a mass spectrometry approach. We collected scleral samples of enucleated eyes due to
Expression of filaggrin-2 protein in the epidermis of human skin diseases: a comparative analysis with filaggrin.
Makino T, et al.
Biochemical and Biophysical Research Communications, 449(1), 100-106 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service