Skip to Content
Merck
All Photos(7)

Key Documents

HPA026813

Sigma-Aldrich

Anti-SPINK2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Acrosin-trypsin inhibitor, Anti-HUSI-II, Anti-Serine protease inhibitor Kazal-type 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Check Cart for Availability


Select a Size

Change View
100 μL
$952.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$952.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

KYRTPNCSQYRLPGCPRHFNPVCGSDMSTYANECTLCMKIREGGHNIKIIRNGPC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SPINK2(6691)

General description

Serine protease inhibitor Kazal-type 2 (SPINK2), also called as HUSI-II (human seminal plasma inhibitor II), is a member of SPINK family, encoded by the gene mapped to human chromosome 4. It is expressed primarily in testis and seminal vesicle. The SPINK2 protein is characterized with a typical kazal domain with six cysteine residues forming three disulfide bridges.

Immunogen

Serine protease inhibitor Kazal-type 2 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Serine protease inhibitor Kazal-type 2 (SPINK2) plays a vital role in tumor progression and response to treatment. Apart from B-lymphoblast TK-6 cells, SPINK2 is highly expressed in most of the leukemia cell lines. Human SPINK2 acts as a trypsin/acrosin inhibitor.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70767

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mapping human haematopoietic stem cells from haemogenic endothelium to birth.
Calvanese, et al.
Nature, 604, 534-540 (2023)
Identification of trypsin-inhibitory site and structure determination of human SPINK2 serine proteinase inhibitor.
Proteins: Structure, Function, and Genetics, 77(1), 209-219 (2009)
Organization and sequence of the gene encoding the human acrosin-trypsin inhibitor (HUSI-II).
Moritz A
Gene, 123(2), 277-281 (1993)
Zine-Eddine Kherraf et al.
EMBO molecular medicine, 9(8), 1132-1149 (2017-05-31)
Azoospermia, characterized by the absence of spermatozoa in the ejaculate, is a common cause of male infertility with a poorly characterized etiology. Exome sequencing analysis of two azoospermic brothers allowed the identification of a homozygous splice mutation in
Vincenza Barresi et al.
iScience, 26(6), 106949-106949 (2023-06-28)
Protease temporary inhibitors are true substrates that bind the catalytic site with high affinity but are slowly degraded, thus acting as inhibitor for a defined time window. Serine peptidase inhibitor Kazal type (SPINK) family is endowed with such functional property

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service