Skip to Content
Merck
All Photos(3)

Key Documents

HPA026294

Sigma-Aldrich

Anti-PRKCH antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PKC-L, Anti-PKCL, Anti-PRKCL

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Check Cart for Availability


Select a Size

Change View
100 μL
$952.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$952.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

FKEIDWAQLNHRQIEPPFRPRIKSREDVSNFDPDFIKEEPVLTPIDEGHLPMINQDEFRNF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRKCH(5583)

General description

Protein kinase C η (PKCL) is expressed in epithelial tissues. It is part of the protein kinase C family. The protein is localized at the Golgi apparatus, nuclear envelope and endoplasmic reticulum. The gene encoding PKCL is located on human chromosome 14q22-q23.

Immunogen

protein kinase C, eta recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Protein kinase C η (PKCL) has a role in differentiation and proliferation of cells. The protein activates IκB kinase (IKK) and modulates nuclear factor κB (NF-κB) upstream signaling. It also has a role in the transcriptional activation of the transglutaminase 1 gene.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78007

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The SNP (rs2230500) in PRKCH decreases the risk of carotid intima-media thickness in a Chinese young adult population.
Wu L
PLoS ONE, 7(7), e40606-e40606 (2012)
Adva Maissel et al.
Cellular signalling, 18(8), 1127-1139 (2005-10-26)
Protein kinase C (PKC) represents a family of serin/threonine kinases, playing a central role in the regulation of cell growth, differentiation and transformation. These enzymes differ in their primary structure, biochemical properties, tissue distribution and subcellular localization. The specific cellular
Protein kinase C? activates NF-?B in response to camptothecin-induced DNA damage.
Raveh-Amit H
Biochemical and Biophysical Research Communications, 412(2), 313-317 (2011)
The eta isoform of protein kinase C mediates transcriptional activation of the human transglutaminase 1 gene.
Ueda E
The Journal of Biological Chemistry, 271(16), 9790-9794 (1996)
Ga Bin Park et al.
Cancer letters, 350(1-2), 5-14 (2014-05-03)
PKCη is involved in proliferation, differentiation, and drug resistance. However, PKCη function in EBV(+) B lymphoma remains poorly understood. Gene silencing of PKCη through siRNA knockdown inhibited cellular proliferation, induced cell cycle arrest in G0/G1 and G2/M phases, and sensitized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service