Skip to Content
Merck
All Photos(2)

Key Documents

HPA025073

Sigma-Aldrich

Anti-SPRTN antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C1orf124, Anti-Zinc finger RAD18 domain-containing protein C1orf124

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Check Cart for Availability


Select a Size

Change View
100 μL
$952.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$952.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL

immunogen sequence

SHQNVLSNYFPRVSFANQKAFRGVNGSPRISVTVGNIPKNSVSSSSQRRVSSSKISLRNSSKVTESASVMPSQDVSGSEDT

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

C1orf124/Spartan, a protein that contains a PCNA-interacting peptide motif, called a PIP box, and a UBZ4 ubiquitin-binding domain, is involved in Rad18-mediated Proliferating Cell Nuclear Antigen (PCNA) ubiquitination and the regulation of DNA damage tolerance via post-replication repair (PRR). It promotes a feed-forward loop to enhance PCNA ubiquitylation and translesion DNA synthesis. Spartan plays an important role in preventing mutations associated with replication of damaged DNA.
Rabbit polyclonal Anti-C1orf124 antibody reacts with human zinc finger RAD18 domain-containing protein C1orf124/spartan.

Immunogen

Zinc finger RAD18 domain-containing protein C1orf124 recombinant protein epitope signature tag (PrEST)

Application

Rabbit polyclonal Anti-C1orf124 antibody is used to tag zinc finger RAD18 domain-containing protein/Spartan for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques such as ELISA, immunoblotting, and immunoprecipitation. It is used as a probe to determine the presence and roles of zinc finger RAD18 domain-containing protein/Spartan in the regulation of DNA damage tolerance via post-replication repair (PRR).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76479

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenji Takeda et al.
Cells, 12(24) (2023-12-22)
Advanced glycation end-products (AGEs), formed through glyceraldehyde (GA) as an intermediate in non-enzymatic reactions with intracellular proteins, are cytotoxic and have been implicated in the pathogenesis of various diseases. Despite their significance, the mechanisms underlying the degradation of GA-derived AGEs
Jaime Lopez-Mosqueda et al.
eLife, 5 (2016-11-18)
Ruijs-Aalfs syndrome is a segmental progeroid syndrome resulting from mutations in the SPRTN gene. Cells derived from patients with SPRTN mutations elicit genomic instability and people afflicted with this syndrome developed hepatocellular carcinoma. Here we describe the molecular mechanism by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service