Skip to Content
Merck
All Photos(2)

Key Documents

HPA024671

Sigma-Aldrich

Anti-RHPN1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-rhophilin 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on24 April 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Estimated to ship on24 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

AFSLLRENFSHAPSPDMSAASLCALEQLMMAQAQECVFEGLSPPASMAPQDCLAQLRLAQEAAQVAAEYRLVHRTMAQPPVHDYVPVSWTALVHVKAE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RHPN1(114822)

General description

The gene RHPN1 (rhophilin Rho GTPase binding protein 1) is mapped to human chromosome 8q24.3. The encoded protein belongs to the rhophilin family of Rho-binding proteins. The mRNA is strongly expressed in the testis. The protein has a Rho binding domain and a PDZ (PSD95, Dlg1 and zo-1) domain.

Immunogen

rhophilin 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

In mouse model, RHPN1 (rhophilin Rho GTPase binding protein 1) is required for the maintenance of the glomerular filtration barrier. Additionally, this protein is a regulator of podocyte cytoskeleton architecture.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76316

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Fujita et al.
Journal of cell science, 113 ( Pt 1), 103-112 (1999-12-11)
The small GTPase Rho; functions as a molecular switch that regulates various cellular processes such as cell adhesion, motility, gene expression and cytokinesis. We previously isolated several putative Rho; targets including rhophilin which bound selectively to the GTP-bound form of
Jeremy W Peck et al.
The Journal of biological chemistry, 277(46), 43924-43932 (2002-09-11)
The Rho GTPases regulate the actin cytoskeleton through interactions with various downstream effector molecules. Here we have identified a ubiquitously expressed human RhoA-binding protein, designated Rhophilin-2. Rhophilin-2 shows 40% amino acid similarity to human Rhophilin-1 and contains an N-terminal Rho-binding
Mark A Lal et al.
Journal of the American Society of Nephrology : JASN, 26(3), 647-662 (2014-07-30)
Rhophilin-1 is a Rho GTPase-interacting protein, the biologic function of which is largely unknown. Here, we identify and describe the functional role of Rhophilin-1 as a novel podocyte-specific protein of the kidney glomerulus. Rhophilin-1 knockout mice were phenotypically normal at
Kyung Im Kim et al.
Basic & clinical pharmacology & toxicology, 111(5), 317-324 (2012-06-08)
Copy number variation (CNV) has been reported to be associated with chemotherapy response, which affects disease prognosis. Here, we determined the frequency of genome-wide cytogenetic CNV aberrations in Korean patients with normal karyotype (NK) acute myeloid leukaemia (AML) and tested

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service