Skip to Content
Merck
All Photos(3)

Key Documents

HPA024516

Sigma-Aldrich

Anti-SP6 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Epfn, Anti-KLF14, Anti-Sp6 transcription factor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Check Cart for Availability


Select a Size

Change View
100 μL
$952.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$952.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

technique(s)

immunoblotting: 1:100-1:250
immunohistochemistry: 1:200-1:500

immunogen sequence

AGGQHLLGPPDGAKALEVAAPESQGLDSSLDGAARPKGSRRSVPRSSGQTVCRCPNCLEAERLGAPCGPDGGKKKHLH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SP6(80320)

Related Categories

General description

SP6 (sp6 transcription factor), also called as KLF14 (kruppel-like factor 14) or Epfn (epiprofin) belongs to SP/KLF (kruppel-like factor) transcription factor family. The protein is encoded by maternally inherited intronless gene, mapped to human chromosome 17q21.3-q22.
KLF14 transcription factor is predominantly expressed in fetal tissues and placenta than in adult tissues. The encoded protein consists of three C2H2-type zinc-fingers attached to each other by kruppel-links at the end of the carboxy-terminal domain and transactivation/suppressor domain at the end of the N-terminal domain.

Immunogen

Sp6 transcription factor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SP6 (sp6 transcription factor) might be involved in regulation of transcription process. In mice, SP6 or Epfn (Epiprofin) is involved in epidermal development.
TGF (transforming growth factor)-β treatment upregulates SP6 or KLF14 (Kruppel-like factor 14), which is a vital non-smad protein that potentially represses the transforming growth factor-β receptor II (TGF-βRII). SP6 aids in tooth development by regulating the epithelial cell fate and tooth number.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75965

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Nakamura et al.
The Journal of biological chemistry, 283(8), 4825-4833 (2007-12-25)
In tooth morphogenesis, the dental epithelium and mesenchyme interact reciprocally for growth and differentiation to form the proper number and shapes of teeth. We previously identified epiprofin (Epfn), a gene preferentially expressed in dental epithelia, differentiated ameloblasts, and certain ectodermal
S Scohy et al.
Genomics, 70(1), 93-101 (2000-11-23)
Using the sequence of the SP1 zinc-finger DNA-binding domain as a probe to screen a mouse EST database, we identified two novel members of the SP/XKLF transcription factor family, KLF13 and KLF14. The mouse Klf13 cDNA (1310 bp in length)
Layla Parker-Katiraee et al.
PLoS genetics, 3(5), e65-e65 (2007-05-08)
Imprinted genes are expressed in a parent-of-origin manner and are located in clusters throughout the genome. Aberrations in the expression of imprinted genes on human Chromosome 7 have been suggested to play a role in the etiologies of Russell-Silver Syndrome
Takashi Nakamura et al.
Journal of cell science, 127(Pt 24), 5261-5272 (2014-10-26)
The basal layer of the epidermis contains stem cells and transit amplifying cells that rapidly proliferate and differentiate further into the upper layers of the epidermis. A number of molecules have been identified as regulators of this process, including p63

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service