Skip to Content
Merck
All Photos(1)

Documents

HPA023964

Sigma-Aldrich

Anti-TTC3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Protein DCRR1, Anti-RING finger protein 105, Anti-TPR repeat protein 3, Anti-TPR repeat protein D, Anti-Tetratricopeptide repeat protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

DCPHVDDCVFAAEFMSNDYVRVTQLYCDGVGVQYKDYIQSERNLEFDICSIWCSKPISVLQDYCDAIKINIFWPLLFQHQNSSVISRLHPCVDANNSRASEINLKKLQHLELMEDIVDLAKKVANDS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TTC3(7267)

General description

TTCR (tetratricopeptide repeat protein 3) is one of the main genes present within the Down syndrome critical region (DSCR) on human chromosome 21q22.2. The encoded protein is an E3 ubiquitin ligase, composed of 2025 amino acids, and its N-terminal contains three tetratricopeptide repeat (TPR) motifs. It also contains a RING (really interesting new gene) finger motif, a putative Akt phosphorylation site, and nuclear localization signals.

Immunogen

Tetratricopeptide repeat protein 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TTCR (tetratricopeptide repeat protein 3) interacts with phosphorylated Akt protein, and promotes its ubiquitination and degradation in the nucleus. The expression of TTCR is increased in Down syndrome (DS) cells, and its interaction with Akt protein is thought to be responsible for the clinical symptoms of DS. TTC3-RhoA-CIT-K (citron kinase) pathway is thought to play a key role in neuronal development, and its hyperactivity might result in disruption of normal differentiation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73898

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Futoshi Suizu et al.
Developmental cell, 17(6), 800-810 (2010-01-12)
The serine threonine kinase Akt is a core survival factor that underlies a variety of human diseases. Although regulatory phosphorylation and dephosphorylation have been well documented, the other posttranslational mechanisms that modulate Akt activity remain unclear. We show here that
Gaia Berto et al.
Journal of cell science, 120(Pt 11), 1859-1867 (2007-05-10)
The Down syndrome critical region (DSCR) on Chromosome 21 contains many genes whose duplication may lead to the major phenotypic features of Down syndrome and especially the associated mental retardation. However, the functions of DSCR genes are mostly unknown and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service