Skip to Content
Merck
All Photos(1)

Documents

HPA023362

Sigma-Aldrich

Anti-GGT6 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GGT 6, Anti-Gamma-glutamyltransferase 6, Anti-Gamma-glutamyltranspeptidase 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

DLAGDALLSLLAGDLGVEVPSAVPRPTLEPAEQLPVPQGILFTTPSPSAGPELLALLEAALRSGAPIPDPCPPFLQTAVSPESSALAAVDSSGSVLLLTSSLNCSFGSAHL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GGT6(124975)

General description

The γ-glutamyltransferase (GGT) enzyme is located in the plasma membrane, with its catalytic site exposed to the extracellular space. It is broadly expressed with most abundant presence in kidney and liver. The gene GGT6 is mapped to human chromosome 17p13.2 and it encodes an isozyme of γ-glutamyltransferase.

Immunogen

Gamma-glutamyltransferase 6 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene GGT6 (γ-glutamyltransferase 6) encodes a member of the γ-glutamyltransferase family of proteins. The glycoprotein GGT catalyzes the extracellular catabolism of glutathione (GSH, γ-glutamyl-cisteinyl-glycine), the crucial thiol intracellular antioxidant compound present in mammalian cells, into glutamic acid and cysteinylglycine. It also functions as a transpeptidase, catalyzing the transfer of the γ-glutamyl moiety to acceptor amino acids. These amino acids assimilate and get reutilized by the cells for GSH synthesis. In the presence of iron or other transition metals, GGT generates reactive oxygen species. This GGT-mediated oxidative stress may be involved in inducing lipid oxidation, oxidation of protein thiols, and activation of certain transcription factors.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75774

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Is serum gamma glutamyltransferase a marker of oxidative stress?
Lee DH
Free Radical Research, 38, 535-539 (2004)
Gamma-glutamyltransferase, atherosclerosis, and cardiovascular disease: triggering oxidative stress within the plaque.
Emdin M
Circulation, 112, 2078-2080 (2005)
Gamma-glutamyltransferase and its isoenzymes: progress and problems.
Nemesanszky E and Lott JA
Clinical Chemistry, 31, 797-803 (1985)
Significance of Polymorphisms and Expression of Enzyme-Encoding Genes Related to Glutathione in Hematopoietic Cancers and Solid Tumors.
Zmorzynski S
BioMed Research International (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service