Skip to Content
Merck
All Photos(3)

Key Documents

HPA021815

Sigma-Aldrich

Anti-ABCF2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-ATP-binding cassette sub-family F member 2, Anti-Iron-inhibited ABC transporter 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

NHLDIETIDALADAINEFEGGMMLVSHDFRLIQQVAQEIWVCEKQTITKWPGDILAYKEHLKSKLVDEEPQLTKRTHNVCTLTLASLPRP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABCF2(10061)

General description

ATP binding cassette subfamily F member 2 (ABCF2) is a cytosolic transporter protein belonging to the ABC (ATP-binding cassette) transporter superfamily. It is expressed in the cytoplasm. The gene encoding ABCF2, consists of a pair of nucleotide-binding domains, and is mapped on human chromosome 7q34-36.

Immunogen

ATP-binding cassette sub-family F member 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ATP binding cassette subfamily F member 2 (ABCF2) has been shown to bind to α-actinin-4 (ACTN4). It suppresses the activity of volume-sensitive outwardly rectifying anion channel (VSOR), which takes part in the efflux of chloride ions in epithelial cells. It has been reported that ABCF2 may be a prognostic marker for ovarian clear cell ovarian adenocarcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74857

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroshi Tsuda et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 11(19 Pt 1), 6880-6888 (2005-10-06)
Patients with ovarian clear cell adenocarcinoma generally have a poor response to combination chemotherapy and have overall poorer prognosis than patients with other histologic types of ovarian cancer. Genetic changes in this group of cancer have not been thoroughly explored.
S Nishimura et al.
British journal of cancer, 99(10), 1651-1655 (2008-11-13)
Uterine cervical and endometrial cancers are common malignant solid neoplasms for which there are no useful prognostic markers. In this study, we evaluate the relationship between ATP-binding cassette superfamily F2 (ABCF2) expression and clinical factors including clinical stage, histologic type
Yuhko Ando-Akatsuka et al.
Journal of cellular physiology, 227(10), 3498-3510 (2012-01-19)
After osmotic swelling, cell volume is regulated by a process called regulatory volume decrease (RVD). Although actin cytoskeletons are known to play a regulatory role in RVD, it is not clear how actin-binding proteins are involved in the RVD process.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service