Skip to Content
Merck
All Photos(6)

Key Documents

HPA021669

Sigma-Aldrich

Anti-CCM2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Cerebral cavernous malformations 2 protein, Anti-Malcavernin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on14 April 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Estimated to ship on14 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

GKKGKKPGIVSPFKRVFLKGEKSRDKKAHEKVTERRPLHTVVLSLPERVEPDRLLSDYIEKEVKYLGQLTSIPGYLNPSSR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCM2(83605)

General description

Cerebral cavernous malformation 2 (CCM2) is an adaptor protein consisting of two domains. Structurally, it has two domains, N-terminal phospho­tyrosine-binding (PTB) domain and an independent domain termed as Karet domain at the C-terminal end. It is mapped on human chromosome 7p. Its phosphotyrosine-binding (PTB) domain has the ability to bind proteins. It is expressed in vascular endothelium and neuroglial precursor cells during development and also observed in arterial endothelium, neurons, some of the glial cells in adult neocortex.

Immunogen

Malcavernin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Cerebral cavernous malformation 2 (CCM2) is associated with several cellular pathways. It mainly behaves as a scaffold protein in the activation of GTPase Rac-dependent p38 mitogen-activated protein kinase (MAPK) during hyperosmotic stress. It also participates in vascular integrity. CCM2 may play a role in vasculogenesis and angiogenesis during the development of the brain. Mutation in the gene is associated with familial cerebral cavernous malformations.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74994

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Denier et al.
American journal of human genetics, 74(2), 326-337 (2004-01-24)
Cerebral cavernous malformations (CCM) are hamartomatous vascular malformations characterized by abnormally enlarged capillary cavities without intervening brain parenchyma. They cause seizures and focal neurological deficits due to cerebral hemorrhages. CCM loci have already been assigned to chromosomes 7q (CCM1), 7p
Rosalia D'Angelo et al.
Gene, 519(1), 202-207 (2012-09-25)
Cerebral cavernous malformations (CCMs; OMIM 116860) are vascular anomalies mostly located in the central nervous system (CNS) and occasionally within the skin and retina. Main clinical manifestations are seizure, hemorrhage, recurrent headaches, focal neurological deficits and epileptic attacks. The CCMs
Xiaoyan Wang et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 68(Pt 6), 683-686 (2012-06-12)
Cerebral cavernous malformation 2 (CCM2) is a novel two-domain adaptor protein which participates in multiple cellular signalling pathways. Loss-of-function mutations in the gene encoding CCM2 are the cause of common human vascular lesions called cerebral cavernous malformations. Here, the purification
Gamze Tanriover et al.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience, 29(5), 509-514 (2011-05-17)
Cerebral cavernous malformation (CCM) is one of the most common types of vascular malformations of the central nervous system, affecting nearly one in 200 people. CCM lesions are characterized by grossly dilated vascular channels lined by a single layer of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service