Skip to Content
Merck
All Photos(3)

Key Documents

HPA021083

Sigma-Aldrich

Anti-SYCP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CT8, Anti-Cancer/testis antigen 8, Anti-SCP-1, Anti-Synaptonemal complex protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

TLGGDSTFFKSFNKCTEDDFEFPFAKTNLSKNGENIDSDPALQKVNFLPVLEQVGNSDCHYQEGLKDSDLENSEGLSRVYSKLYKEAEKIKKWKVSTEAELRQKESKLQENRKIIEAQRKAIQEL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYCP1(6847)

General description

The gene SYCP1 (synaptonemal complex protein 1) is mapped to human chromosome 1p13-p12.

Immunogen

Synaptonemal complex protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SYCP1 (synaptonemal complex protein 1) regulates meiosis during gametogenesis. In meiosis, the synaptonemal complex contains two lateral elements (LEs) joined by transverse filaments. SYCP1 is a component of transverse filaments and plays a crucial role in connecting gap between the LE and CE (central element). It is up-regulated in cutaneous T-cell lymphoma (CTCL) and some nasopharyngeal carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74749

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivan V Litvinov et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(14), 3799-3808 (2014-05-23)
The pathogenesis of cutaneous T-cell lymphoma (CTCL) remains only partially understood. A number of recent studies attempted to identify novel diagnostic markers and future therapeutic targets. One group of antigens, cancer-testis (CT) antigens, normally present solely in testicular germ cells
Yael Costa et al.
Journal of cell science, 118(Pt 12), 2755-2762 (2005-06-10)
Completion of meiosis in mammals depends on the formation of the synaptonemal complex, a tripartite structure that physically links homologous chromosomes during prophase I. Several components of the synaptonemal complex are known, including constituents of the cohesin core, the axial/lateral
Min Yi et al.
Lin chuang er bi yan hou tou jing wai ke za zhi = Journal of clinical otorhinolaryngology, head, and neck surgery, 21(8), 343-345 (2007-07-05)
To investigate the gene expression of Cancer-Testis Antigen SCP-1 in human nasopharyngeal carcinoma. The SCP-1 mRNA were examined in the tumor tissues of 28 nasopharyngeal carcinoma cases and control pharynx vasitis tissues of 40 non-cancer cases by reverse transcriptase polymerase
Koji Okudela et al.
Pathology international, 60(10), 651-660 (2010-09-18)
Lung cancer, like other cancers, is considered to develop through the accumulation of genetic alterations. Mutation of the KRAS gene is one of the most important events in carcinogenesis of the lung. The KRAS gene, belonging to the RAS gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service