Skip to Content
Merck
All Photos(3)

Key Documents

HPA020914

Sigma-Aldrich

Anti-METTL1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C12orf1, Anti-TRM8, Anti-TRMT8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on03 May 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$1,040.00


Estimated to ship on03 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

MCTHFEEHPLFERVPLEDLSEDPVVGHLGTSTEEGKKVLRNGGKNFPAIFRRIQDPVLQAVTSQTSLPGH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... METTL1(4234)

General description

The gene METTL1 (methyltransferase-like protein 1) is mapped to human chromosome 12q13. It is ubiquitously expressed. METTL1 is a mammaliam ortholog of yeast Trm8 (transfer RNA methyltransferase 8).[1]

Immunogen

tRNA recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

In yeast, Trm8 (transfer RNA methyltransferase 8) is crucial for protecting tRNA from rapid tRNA decay.[1] In yeast extract, human METTL1 (methyltransferase-like protein 1) shows m7G-methyltransferase activity. METTL1 is up-regulated in many cancers.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74730

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiu-Hong Tian et al.
Journal of molecular medicine (Berlin, Germany), 97(11), 1535-1545 (2019-08-30)
RNA methylation is emerging as an important regulator of gene expression. Dysregulation of methyltransferase that is essential for RNA modification contributes to the development and progression of human cancers. Here we show that methyltransferase-like 1 (METTL1) is upregulated in hepatocellular
A Bahr et al.
Genomics, 57(3), 424-428 (1999-05-18)
A novel human gene, METTL1, has been identified by its sequence similarity to the yeast ORF YDL201w. The human cDNA and the genomic structure of METTL1 have been analyzed. The transcript contains 1292 nucleotides and codes for a protein of
Andrei Alexandrov et al.
RNA (New York, N.Y.), 8(10), 1253-1266 (2002-10-31)
7-methylguanosine (m7G) modification of tRNA occurs widely in eukaryotes and bacteria, is nearly always found at position 46, and is one of the few modifications that confers a positive charge to the base. Screening of a Saccharomyces cerevisiae genomic library
Mayumi Okamoto et al.
PLoS genetics, 10(9), e1004639-e1004639 (2014-09-19)
Nonessential tRNA modifications by methyltransferases are evolutionarily conserved and have been reported to stabilize mature tRNA molecules and prevent rapid tRNA decay (RTD). The tRNA modifying enzymes, NSUN2 and METTL1, are mammalian orthologs of yeast Trm4 and Trm8, which are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service