Skip to Content
Merck
All Photos(4)

Key Documents

HPA020880

Sigma-Aldrich

Anti-DPF2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BAF45d, Anti-REQ, Anti-ubi-d4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

SFPSIKPDTDQTLKKEGLISQDGSSLEALLRTDPLEKRGAPDPRVDDDSLGEFPVTNSRARKRILEPDDFLDDLDDEDYEEDTPKRRGKGKSKGKGVGS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DPF2(5977)

General description

DPF2 (D4, zinc and double PHD fingers family 2) belongs to the d4-protein family. The protein contains an amino-terminal 2/3 domain, a C2H2-type zinc finger and a carboxyl-terminal PHD (plant homeodomain) zinc finger. It is also referred to as REQ (requiem).

Immunogen

Zinc finger protein ubi-d4 recombinant protein epitope signature tag (PrEST)

Application

Anti-DPF2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting.[1] To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

DPF2 (D4, zinc and double PHD fingers family 2) is a transcription factor. It is also identified as an early apoptosis-signaling gene. DPF2 functions as an adaptor protein between the SWI/SNF (SWItch/Sucrose Non-Fermentable) complex and RELB (v-rel avian reticuloendotheliosis viral oncogene homolog B)/p52, and thereby promotes the NF (nuclear factor)-κB-dependent transcriptional activation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73709

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshio Tando et al.
The Journal of biological chemistry, 285(29), 21951-21960 (2010-05-13)
The SWI/SNF chromatin remodeling complex plays pivotal roles in mammalian transcriptional regulation. In this study, we identify the human requiem protein (REQ/DPF2) as an adaptor molecule that links the NF-kappaB and SWI/SNF chromatin remodeling factor. Through in vitro binding experiments
Wei Zhang et al.
Biochemical and biophysical research communications, 413(1), 58-61 (2011-09-06)
DPF2 is an evolutionary highly conserved member of the d4-protein family characterized by an N-terminal 2/3 domain, a C2H2-type zinc finger (ZF), and a C-terminal tandem PHD zinc finger. DPF2 is identified as a transcription factor and may be related
V Adam Cruickshank et al.
PloS one, 10(11), e0142806-e0142806 (2015-11-17)
Alterations in chromatin structure caused by deregulated epigenetic mechanisms collaborate with underlying genetic lesions to promote cancer. SMARCA4/BRG1, a core component of the SWI/SNF ATP-dependent chromatin-remodelling complex, has been implicated by its mutational spectrum as exerting a tumour-suppressor function in
Yiping Lim et al.
Molecular biotechnology, 46(3), 301-307 (2010-06-24)
Apoptosis is known to be the main cause of cell death in the bioreactor environment, leading to the loss of recombinant protein productivity. In a previous study, transcriptional profiling was used to identify and target four early apoptosis-signaling genes: FADD

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service