Skip to Content
Merck
All Photos(3)

Key Documents

HPA020879

Sigma-Aldrich

Anti-ZMYM5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MYM, Anti-ZNF198L1, Anti-ZNF237, Anti-zinc finger, MYM-type 5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

KEKTSQLQLSVECGTDTLLIQENVNLPPSSTSTIADTFQEQLEEKNFEDSIVPVVLSADPGTWPRILNIKQRDTLVENVP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZMYM5(9205)

Related Categories

General description

The gene ZMYM5 (zinc finger MYM-type protein 5) is mapped to human chromosome 13q11-q12. It belongs to the MYM gene family. ZMYM5 is also referred to as ZNF237 (zinc finger protein 237).

Immunogen

zinc finger, MYM-type 5 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ZMYM5 (zinc finger MYM-type protein 5) is involved in transcriptional modulation. It regulates expression of presenilin 1, and thereby suppresses presenilin 1-controlled endoplasmic reticulum Ca2+ leak channels.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73708

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hriday K Das et al.
Journal of neurochemistry, 122(3), 487-500 (2012-05-23)
Genetic deletion or mutations of presenilin genes (PS1/PS2) cause familial Alzheimer's disease and calcium (Ca²⁺) signaling abnormalities. PS1/PS2 act as endoplasmic reticulum (ER) Ca²⁺ leak channels that facilitate passive Ca²⁺ leak across ER membrane. Studies with PS1/PS2 double knockout (PS1/PS2-DKO)
Martine Pastorcic et al.
Brain research, 1128(1), 21-32 (2006-11-28)
DNA sequences required for the expression of the human presenilin 1 (PS1) gene have been identified between -118 and +178 flanking the major initiation site (+1) mapped in SK-N-SH cells. Several Ets sites are located both upstream as well as
J Sohal et al.
Cytogenetics and cell genetics, 89(1-2), 24-28 (2000-07-15)
We have cloned a novel, widely expressed human gene, ZNF237, that shows extensive similarity to the N-terminal region of ZNF198. Two alternatively spliced regions were identified by RT-PCR; the major splice variant is predicted to encode a 383 amino acid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service