Skip to Content
Merck
All Photos(3)

Key Documents

HPA019938

Sigma-Aldrich

Anti-DFFA antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-DFF-45, Anti-DNA fragmentation factor 45 kDa subunit, Anti-DNA fragmentation factor subunit alpha, Anti-ICAD, Anti-Inhibitor of CAD

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Estimated to ship on27 April 2025



Select a Size

Change View
100 μL
$952.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$952.00


Estimated to ship on27 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

LRQSCAIVQRLQHTLQQVLDQREEVRQSKQLLQLYLQALEKEGSLLSKQEESKAAFGEEVDAVDTGISRETSSDVALASHILTAL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DFFA(1676)

General description

DFFA (DNA fragmentation factor, alpha polypeptide) is a heterodimeric 45kDa protein mapped on chromosome location 1p36.2-36.3. Its expression has also been found in neuroblastoma (NB) cell lines.[1]

Immunogen

DNA fragmentation factor subunit alpha recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

DFFA (DNA fragmentation factor, α polypeptide) functions as a chaperone of DNA fragmentation factor 40 (DFF40) in apoptosis and balancing genomic stability. The activity of DFF is induced by cleavage of DFFA by caspase-3, resulting in release of DFF40, which degrades chromosomal DNA. During menstruation, the apoptosis of endometrium cells is associated with DFFA activation. DFFA expression decreases after menopause. DFFA is a target of miR-145.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74220

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youssef Errami et al.
PloS one, 8(2), e57871-e57871 (2013-03-02)
We previously showed that DNA fragmentation factor, which comprises a caspase-3-activated DNase (CAD) and its inhibitor (ICAD), may influence the rate of cell death by generating PARP-1-activating DNA breaks. Here we tested the hypothesis that ICAD-deficient colon epithelial cells exhibiting
Tomasz Banas et al.
Gynecologic and obstetric investigation, 73(3), 177-182 (2012-03-02)
Human endometrium undergoes cyclic structural and functional modifications, and if no conception occurs menstruation is observed as the result of endometrial cell apoptosis via DFF40/DFF45 complex activation. In postmenopausal endometrium, the proliferative potential of endometrial cells is decreased, while their
Yiannis P Ninios et al.
Apoptosis : an international journal on programmed cell death, 15(2), 128-138 (2009-11-03)
A major hallmark of the terminal stages of apoptosis is the internucleosomal DNA fragmentation. The endonuclease responsible for this type of DNA degradation is the DNA fragmentation factor (DFF). DFF is a complex of the endonuclease DFF40 and its chaperone/inhibitor
Jianjun Zhang et al.
Molecular cancer, 9, 211-211 (2010-08-07)
MicroRNA-145 (miR-145) is considered to play key roles in many cellular processes, such as proliferation, differentiation and apoptosis, by inhibiting target gene expression. DNA Fragmentation Factor-45 (DFF45) has been found to be the substrate of Caspase-3, and the cleavage of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service