Skip to Content
Merck
All Photos(2)

Key Documents

HPA019823

Sigma-Aldrich

Anti-FAM20C antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DMP-4, Anti-Dentin matrix protein 4, Anti-Protein FAM20C

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$1,040.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

immunogen sequence

VNSDTRLSPKAAENPDWPHAGAEGAEFLSPGEAAVDSYPNWLKFHIGINRYELYSRHNPAIEALLHDLSSQRITSVAMKSGGTQLKLIM

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FAM20C(56975)

General description

FAM20C (Family with sequence similarity 20, member C) is a member of FAM20 protein family. It is distributed in several mammalian cell lines. During hematopoietic differentiation, it is expressed in hematopoietic cells.

Immunogen

Dentin matrix protein 4 Precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-FAM20C antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues.[1] These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

FAM20C (Family with sequence similarity 20, member C) is a secretory Golgi casein kinase involved in biomineralization, enamel formation, lipid homeostasis, wound healing, cell migration and adhesion. It has ability to phosphorylate S-x-E/pS motifs on proteins in milk and in the extracellular matrix of bones and teeth. During enamel formation, it forms a functional complex by binding to a pseudokinase, Fam20A, which triggers extracellular protein phosphorylation within the secretory pathway. Mutations in FAM20C gene cause Raine syndrome, hypophosphatemia, hyperphosphaturia, dental anomalies, intracerebral calcifications and osteosclerosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75083

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shih-Kai Wang et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 28(12), 2508-2511 (2013-05-25)
FAM20C, also known as Golgi casein kinase (G-CK), is proposed to be the archetype for a family of secreted kinases that phosphorylate target proteins in the Golgi and in extracellular matrices, but FAM20C serving an extracellular function is controversial. FAM20C
Hypophosphatemic osteomalacia and bone sclerosis caused by a novel homozygous mutation of the FAM20C gene in an elderly man with a mild variant of Raine syndrome.
Takeyari S, et al.
Bone, 67, 56-62 (2014)
Vincent S Tagliabracci et al.
Cell, 161(7), 1619-1632 (2015-06-20)
The existence of extracellular phosphoproteins has been acknowledged for over a century. However, research in this area has been undeveloped largely because the kinases that phosphorylate secreted proteins have escaped identification. Fam20C is a kinase that phosphorylates S-x-E/pS motifs on
Jixin Cui et al.
eLife, 4, e06120-e06120 (2015-03-20)
Although numerous extracellular phosphoproteins have been identified, the protein kinases within the secretory pathway have only recently been discovered, and their regulation is virtually unexplored. Fam20C is the physiological Golgi casein kinase, which phosphorylates many secreted proteins and is critical
Exome sequencing reveals FAM20c mutations associated with fibroblast growth factor 23-related hypophosphatemia, dental anomalies, and ectopic calcification.
Rafaelsen SH, et al.
Bone and Mineral, 28, 1378-1385 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service