Skip to Content
Merck
All Photos(4)

Key Documents

HPA019545

Sigma-Aldrich

Anti-PARL antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Mitochondrial intramembrane cleaving protease PARL, Anti-Pbeta, Anti-Presenilins-associated rhomboid-like protein, mitochondrial

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

ESLKSRVQSYFDGIKADWLDSIRPQKEGDFRKEINKWWNNLSDGQR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PARL(55486)

General description

PARL (Presenilin associated, rhomboid-like) is a nuclear modifier gene expressed in both outer and inner mitochondrial membranes.

Immunogen

presenilin associated rhomboid like recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PARL (Presenilin associated, rhomboid-like) plays a key role in the apoptotic regulation during postnatal growth. It controls activity of an antiapoptotic form of OPA1, which prevents the release of mitochondrial cytochrome c in response to intrinsic apoptoptic signals, through proteolytic processing.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73166

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rochmy Istikharah et al.
Experimental eye research, 116, 55-57 (2013-08-27)
The present study explored variation in the PARL gene as one of the potential nuclear modifiers in the pathogenesis of Leber hereditary optic neuropathy (LHON). Ten exons, their franking introns and 3' UTR of the PARL gene were analysed. Seventeen
Attila Sík et al.
The Journal of biological chemistry, 279(15), 15323-15329 (2004-01-21)
Regulated intramembrane proteolysis (RIP) is an emerging paradigm in signal transduction. RIP is mediated by intramembrane-cleaving proteases (I-CliPs), which liberate biologically active nuclear or secreted domains from their membrane-tethered precursor proteins. The yeast Pcp1p/Rbd1p protein is a Rhomboid-like I-CliP that
Danny V Jeyaraju et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(49), 18562-18567 (2006-11-23)
Remodeling of mitochondria is a dynamic process coordinated by fusion and fission of the inner and outer membranes of the organelle, mediated by a set of conserved proteins. In metazoans, the molecular mechanism behind mitochondrial morphology has been recruited to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service