Skip to Content
Merck
All Photos(5)

Key Documents

HPA019486

Sigma-Aldrich

Anti-ING2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ING1Lp, Anti-Inhibitor of growth 1-like protein, Anti-Inhibitor of growth protein 2, Anti-p32, Anti-p33ING2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

TQMLELVENRARQMELHSQCFQDPAESERASDKAKMDSSQPERSSRRPRRQRTSESRDLCHMANGIEDCDDQPPKEKKSKSAKK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ING2(3622)

General description

ING2 (Inhibitor of growth family, member 2) is a component of a chromatin-regulatory complex belonging to lNG family. It is composed of three exons, 1a, 1b, and 2.[1]

Immunogen

Inhibitor of growth protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ING2 (Inhibitor of growth family, member 2) acts as a tumor suppressor gene in osteosarcoma, basal cell carcinoma (BCC) by controlling cell cycle progression and cell proliferation. It is associated with several cellular activities such as cell cycle regulation, DNA repair, apoptosis and senescence. It also performs in chromatin remodeling and transcriptional regulation by histone modification.[1][2][1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74000

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Rui Han et al.
Oncology reports, 32(5), 1967-1972 (2014-09-06)
Osteosarcoma is a high-grade malignant bone tumor. Loss of inhibitor of growth 2 (ING2) expression has been demonstrated in numerous types of cancers. However, no study has shown the relationship between ING2 expression and osteosarcoma. In the present study, we confirmed
Dennis J Bua et al.
Scientific reports, 3, 2137-2137 (2013-07-05)
ING2 (inhibitor of growth family member 2) is a component of a chromatin-regulatory complex that represses gene expression and is implicated in cellular processes that promote tumor suppression. However, few direct genomic targets of ING2 have been identified and the
Rebecca J Harris et al.
Nature communications, 14(1), 7243-7243 (2023-11-10)
Histone modifications influence the recruitment of reader proteins to chromosomes to regulate events including transcription and cell division. The idea of a histone code, where combinations of modifications specify unique downstream functions, is widely accepted and can be demonstrated in
Y Q Pan et al.
Neoplasma, 61(4), 468-475 (2014-04-10)
The inhibitor of growth 2 (ING2) is a member of lNG family, involved in cell cycle regulation, DNA repair, apoptosis and senescence, and participating in chromatin remodeling and transcriptional regulation by histone modification. Recent researches suggest ING2 plays roles in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service