Skip to Content
Merck
All Photos(2)

Key Documents

HPA019062

Sigma-Aldrich

Anti-NEK3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HSPK36, Anti-MGC29949, Anti-NIMA (never in mitosis gene a)-related kinase 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

NLILKVCQGCISPLPSHYSYELQFLVKQMFKRNPSHRPSATTLLSRGIVARLVQKCLPPEIIMEYGEEVLEEIKNSKHNTPRKKTNPSRIRIALGNEASTV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEK3(4752)

General description

The gene NEK3 (never in mitosis A-related kinase 3) is mapped to human chromosome 13q14.13. It belongs to NIMA (never in mitosis A) family of proteins. It is strongly expressed in testis, ovary, and brain.

Immunogen

NIMA (never in mitosis gene a)-related kinase 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PRL (prolactin)-mediated association between NEK3 (never in mitosis A-related kinase 3) and VAV1/2 (vav guanine nucleotide exchange factor) is crucial for PRL-induced cytoskeletal re-organization and motility of breast cancer cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73489

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Hernández et al.
Cancer investigation, 24(7), 682-688 (2006-11-23)
Chromosomal region 13q14 is frequently deleted in prostate cancer. nek3, a protein kinase related gene, is located on this region. Analysis of the coding region of nek3 showed an A insertion/deletion polymorphism in a stretch of adenines at the end
Sommer L Miller et al.
Molecular endocrinology (Baltimore, Md.), 19(4), 939-949 (2004-12-25)
Prolactin (PRL) receptor activation contributes to the progression and motility of human breast cancer. This event activates multimeric signaling pathways, including the activation of the Vav family of guanine nucleotide exchange factors. To detect novel proteins interacting with Vav, yeast
M Kimura et al.
Cytogenetics and cell genetics, 95(3-4), 177-182 (2002-06-14)
NEKs (NIMA-related kinases) are a group of protein kinases sharing high amino acid sequence identities with NIMA (never in mitosis gene a) which control mitosis in Aspergillus nidulans. We have cloned a cDNA for human NEK3, a novel human gene
S L Miller et al.
Oncogene, 26(32), 4668-4678 (2007-02-14)
Prolactin (PRL) stimulates the cytoskeletal re-organization and motility of breast cancer cells. During PRL receptor signaling, Vav2 becomes phosphorylated and activated, an event regulated by the serine/threonine kinase Nek3. Given the regulatory role of Vav2, the function of Nek3 in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service