Skip to Content
Merck
All Photos(4)

Key Documents

HPA019060

Sigma-Aldrich

Anti-RAP1GDS1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Exchange factor smgGDS, Anti-Rap1 GTPase-GDP dissociation stimulator 1, Anti-SMG GDS protein, Anti-SMG P21 stimulatory GDP/GTP exchange protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on27 April 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Estimated to ship on27 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

ESSKQFASTNIAEELVKLFKKQIEHDKREMIFEVLAPLAENDAIKLQLVEAGLVECLLEIVQQKVDSDKEDDITELKTGSDLMVLLLLGDESMQKLFEGGKGSVFQRVLSWIPSNNHQLQLAGA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAP1GDS1(5910)

General description

The gene RAP1GDS1 (rap1 GTPase-GDP dissociation stimulator 1) is mapped to human chromosome 4q23. The protein localizes in the cytoplasm, mitochondria and endoplasmic reticulum. RAP1GDS1 is also referred as smgGDS (SMG P21 stimulatory GDP/GTP exchange protein).

Immunogen

Rap1 GTPase-GDP dissociation stimulator 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RAP1GDS1 (rap1 GTPase-GDP dissociation stimulator 1) is a guanine exchange factor which is responsible for promoting prenylation and trafficking of Ras and Rho family members containing polybasic region. The longer form of RAP1GDS1 controls entry of non-prenylated GTPases into the prenylation pathway. The shorter form regulates trafficking of prenylated GTPases to the plasma membrane. RAP1GDS1 also regulates cell cycle progression. It is up-regulated in non-small cell lung carcinoma, prostate cancer, breast cancer and pancreatic cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74807

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew D Hauser et al.
Molecular cancer research : MCR, 12(1), 130-142 (2013-11-08)
Breast cancer malignancy is promoted by the small GTPases RhoA and RhoC. SmgGDS is a guanine nucleotide exchange factor that activates RhoA and RhoC in vitro. We previously reported that two splice variants of SmgGDS, SmgGDS-607, and SmgGDS-558, have different
Nathan J Schuld et al.
Cell cycle (Georgetown, Tex.), 13(6), 941-952 (2014-02-21)
Oncogenic mutation or misregulation of small GTPases in the Ras and Rho families can promote unregulated cell cycle progression in cancer. Post-translational modification by prenylation of these GTPases allows them to signal at the cell membrane. Splice variants of SmgGDS
Mariana Igoillo-Esteve et al.
PLoS genetics, 9(10), e1003888-e1003888 (2013-11-10)
We describe a new syndrome of young onset diabetes, short stature and microcephaly with intellectual disability in a large consanguineous family with three affected children. Linkage analysis and whole exome sequencing were used to identify the causal nonsense mutation, which
Yu-Fan Hsieh et al.
PloS one, 8(12), e81516-e81516 (2013-12-19)
Transglutaminase 2 (TG2) is a protein cross-linking enzyme known to be associated with the in vivo apoptosis program of T cells. However, its role in the T cell apoptosis program was not investigated yet. Here we report that timed overexpression

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service