Skip to Content
Merck
All Photos(8)

Key Documents

HPA018505

Sigma-Aldrich

Anti-LRMP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Lymphoid-restricted membrane protein, Anti-Protein Jaw1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

RSASLNSKPSSLRRVTIASLPRNIGNAGMVAGMENNDRFSRRSSSWRILGSKQSEHRPSLPRFISTYSWADAEEEKCELKTKDDSEPSGEETVERTRKPSLSEKKNNPSKWDVSSVYD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LRMP(4033)

General description

The gene LRMP (lymphoid-restricted membrane protein) is mapped to human chromosome 12p12.1. LRMP transcript is abundantly expressed in pre-B, pre-T, B cell lines and thymocytes. LRMP is also expressed in sweet, bitter, and umami taste receptor-expressing cells of mouse circumvallate, foliate, and fungiform papillae. The protein contains a conserved coiled-coil domain in the middle of the protein and a carboxyl-terminal transmembrane domain. LRMP is localized in the endoplasmic reticulum membrane.

Immunogen

Lymphoid-restricted membrane protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Lymphoid-restricted membrane protein (LRMP) associates with type III IP3 (inositol 1,4,5-triphosphate) receptor (IP3R3) and participate in IP3-Ca2+ signal cascade in sweet, bitter, and umami taste signal transduction. Single nucleotide polymorphism in LRMP is associated with predicting the survival in patients with lung adenocarcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86505

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giacomo Manenti et al.
Carcinogenesis, 27(7), 1386-1390 (2006-01-18)
Mouse Lrmp and Casc1 genes are candidates for the pulmonary adenoma susceptibility 1 (Pas1) locus, the major determinant of strain variation in lung tumor susceptibility. These genes contain coding and non-coding single nucleotide polymorphisms (SNPs) strongly associated with lung tumor
Yoichiro Shindo et al.
Chemical senses, 35(2), 171-177 (2010-01-15)
Inositol 1,4,5-triphosphate-mediated calcium (IP3-Ca2+) signal cascade is an essential process in sweet, bitter, and umami taste signal transduction. Although the main components of this cascade have been identified, the candidate regulators of them in taste tissues are still unclear. In
Stephanie Ma et al.
International journal of cancer, 124(10), 2323-2332 (2009-01-29)
Identification of genomic alterations associated with the progression of prostate cancer may facilitate the better understanding of the development of this highly variable disease. Matched normal, premalignant high-grade prostatic intraepithelial neoplasia and invasive prostate carcinoma cells were procured by laser
T W Behrens et al.
Journal of immunology (Baltimore, Md. : 1950), 153(2), 682-690 (1994-07-15)
Jaw1 is a novel lymphoid-restricted gene that is expressed in a developmentally regulated fashion in both the B and T cell lineages. Jaw1 mRNA is abundantly expressed in pre-B and B cell lines with minimal or undetectable expression in plasma

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service