Skip to Content
Merck
All Photos(3)

Key Documents

HPA018151

Sigma-Aldrich

Anti-CDR2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cerebellar degeneration-related protein 2, Anti-Major Yo paraneoplastic antigen, Anti-Paraneoplastic cerebellar degeneration-associated antigen

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

GVEKLVPDSLYVPFKEPSQSLLEEMFLTVPESHRKPLKRSSSETILSSLAGSDIVKGHEETCIRRAKAVKQRGISLLHEVDTQYSALKVKYEELLKKCQEEQDSLSHKAVQTSRAAAKDLTGV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDR2(1039)

General description

The gene cerebellar degeneration-related protein-2 (CDR2) has been mapped to human chromosome 16p12.3. The gene encodes a cytoplasmic protein harboring a helix-leucine zipper motif. CDR2 is localized in the cytoplasm.[1][2] The mRNA is ubiquitously expressed in adult mouse tissue with most abundance in the testis. However, the protein is present predominantly in brain, testis, gynecological and breast tumors, suggesting post-transcriptional regulation. CDR2 is cell cycle regulated in tumor cells with highest expression in mitosis.[2]

Immunogen

Cerebellar degeneration-related protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Cerebellar-degeneration-related antigen-2 (CDR2) is a target antigen in paraneoplastic cerebellar degeneration (PCD).[2] In PCD, CDR2 has been shown to activate specific cytotoxic T lymphocytes which are likely to contribute to the development of autoimmune neuronal degeneration. CDR2 is up-regulated in breast and ovarian tumor cells obtained from the general population of cancer patients. In addition, it is shown to attenuate hypoxia response in renal cell carcinoma by regulating prolyl-4-hydroxylase domain protein abundance. Through its leucine zipper motif CDR2 interacts with c-myc in adult cerebellar Purkinje neurons, tumor cell lines and cerebellar extracts. CDR2 down-regulates c-myc-dependent transcription. The protein is required for mitosis, as CDR2 knockdown cells have aberrant mitotic spindles and impaired proliferation.[2]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73719

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Darnell et al.
Cancer research, 60(8), 2136-2139 (2000-04-29)
The paraneoplastic neurological disorders provide perhaps the best known example of naturally occurring tumor immunity in humans. For example, patients with paraneoplastic cerebellar degeneration (PCD) appear to suppress the growth of occult breast or ovarian tumors that express a neuronal
K Balamurugan et al.
Oncogene, 28(37), 3274-3285 (2009-07-08)
The onconeuronal cerebellar degeneration-related antigen Cdr2 is associated with paraneoplastic syndromes. Neoplastic expression of Cdr2 in ovary and breast tumors triggers an autoimmune response that suppresses tumor growth by developing tumor immunity, but culminates in cerebellar degeneration when Cdr2-specific immune
M L Albert et al.
Nature medicine, 4(11), 1321-1324 (1998-11-11)
Models for immune-mediated tumor regression in mice have defined an essential role for cytotoxic T lymphocytes (CTLs); however, naturally occurring tumor immunity in humans is poorly understood. Patients with paraneoplastic cerebellar degeneration (PCD) provide an opportunity to explore the mechanisms
Cecilie Totland et al.
Cancer immunology, immunotherapy : CII, 60(2), 283-289 (2010-11-17)
Paraneoplastic cerebellar degeneration (PCD) is often associated with Yo antibodies that are directed against human cerebellar degeneration-related protein 2 (CDR2). Such antibodies may also be found in ovarian cancer patients without PCD. We studied if there was an association between
H J Okano et al.
Genes & development, 13(16), 2087-2097 (1999-08-31)
Paraneoplastic cerebellar degeneration (PCD) is a disorder in which breast or ovarian tumors express an onconeural antigen termed cdr2, which normally is expressed in cerebellar Purkinje neurons. This leads to an immune response to cdr2 that is associated with tumor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service