Skip to Content
Merck
All Photos(6)

Key Documents

HPA015721

Sigma-Aldrich

Anti-B4GALNT2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti- beta- 1,4-GalNAc transferase, Anti-Beta-1,4 N-acetylgalactosaminyltransferase 2, Anti-Sd(a, Anti-UDP-GalNAc:Neu5Aca2-3Galb-R b1,4-N- acetylgalactosaminyltransferase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200
western blot: 0.04-0.4 μg/mL

immunogen sequence

LLPEERLRNLFSYDGIWLFPKNQCKCEANKEQGGYNFQDAYGQSDLPAVKARRQAEFEHFQRREGLPRPLPLLVQPNLPFGYPVHGVEVMPLHTVPIPGLQFEGPDA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Beta-1,4 N-acetylgalactosaminyltransferase 2 recombinant protein epitope signature tag (PrEST)

Application

Anti-B4GALNT2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

β-1,4 N-acetylgalactosaminyltransferase 2 is an enzyme encoded by the B4GALNT2 gene in humans. It helps in stimulating the glycosylation of α dystroglycan and up-regulates synaptic dystroglycan-binding proteins, when overexpressed. It may act as a therapeutic in various muscular dystrophy. This gene is mainly regulated by the promoter DNA methylation. It is also involved in the Sd(a) synthesis, which is useful for decrease in cancer lesions of the gastrointestinal tract.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72178

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Srikar Krishna et al.
EMBO reports, 20(7), e47789-e47789 (2019-07-04)
Transfer RNA (tRNA)-derived small RNAs (tsRNAs) have recently emerged as important regulators of protein translation and shown to have diverse biological functions. However, the underlying cellular and molecular mechanisms of tsRNA function in the context of dynamic cell-state transitions remain
Sjoerd van der Post et al.
Molecular & cellular proteomics : MCP, 13(9), 2277-2287 (2014-06-04)
The colonic epithelium is a highly dynamic system important for the regulation of ion and water homeostasis via absorption and secretion and for the maintenance of a protective barrier between the outer milieu and the inside of the body. These
Hou-Ren Wang et al.
Glycobiology, 18(1), 104-113 (2007-10-30)
It has been noted that the expression of Sd(a), including its antigenic structure, the beta-1,4-N-acetylgalactosyltransferase II (beta4GalNAcT-II) activity responsible for its formation, and the Sd(a) beta4GalNAcT-II mRNA transcript, is drastically reduced in oncogenetic processes in gastrointestinal tissues. Markedly reduced metastatic
Yuki I Kawamura et al.
Cancer research, 65(14), 6220-6227 (2005-07-19)
The Sd(a) blood group carbohydrate structure is expressed in the normal gastrointestinal mucosa. We reported previously that the expression of Sd(a) carbohydrate structures and beta1,4-N-acetylgalactosaminyltransferase (beta1,4GalNAcT) activity responsible for Sd(a) synthesis were remarkably decreased in cancer lesions of the gastrointestinal
Virginie Cogez et al.
International journal of molecular sciences, 24(4) (2023-02-26)
The Sda carbohydrate epitope and its biosynthetic B4GALNT2 enzyme are expressed in the healthy colon and down-regulated to variable extents in colon cancer. The human B4GALNT2 gene drives the expression of a long and a short protein isoform (LF-B4GALNT2 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service