Skip to Content
Merck
All Photos(1)

Key Documents

HPA015641

Sigma-Aldrich

Anti-USP7 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Deubiquitinating enzyme 7, Anti-Herpesvirus-associated ubiquitin-specific protease, Anti-Ubiquitin carboxyl-terminal hydrolase 7, Anti-Ubiquitin thioesterase 7, Anti-Ubiquitin-specific-processing protease 7

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

RSEATFQFTVERFSRLSESVLSPPCFVRNLPWKIMVMPRFYPDRPHQKSVGFFLQCNAESDSTSWSCHAQAVLKIINYRDDEKSFSRRISHLFFHKENDWGFSNFMAWSEVTDPEKGFIDDDKVTFEVFVQADAPHGVAWDSK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USP7(7874)

Immunogen

Ubiquitin carboxyl-terminal hydrolase 7 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Ubiquitin-specific-processing protease 7 (USP7) is an enzyme encoded by the USP7 gene in humans. It is a deubiquitinating enzyme that regulates proteins involved in cell cycle and forms a negative feedback loop in insulin/IGF signaling. It also functions as a tumor suppressor and oncogene. Overexpression of this gene is associated with oncogenic role in glioma progression, may regulate squamous cell carcinoma and large cell carcinoma cells and may act as a therapeutic target for the same. It may also act as a candidate gene for colorectal cancer (CRC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72648

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuandong Cheng et al.
Oncology reports, 29(5), 1730-1736 (2013-03-14)
The human herpesvirus-associated ubiquitin-specific protease (HAUSP) deubiquitinating enzyme has been shown to regulate many proteins involved in the cell cycle, as well as tumor suppressors and oncogenes. However, the expression pattern of HAUSP in glioma patients is still unclear. The
Guang-Yin Zhao et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 36(3), 1721-1729 (2014-12-19)
In non-small cell lung cancer (NSCLC), both USP7 expression and p53 gene status were reported to be an indicator of poor prognosis in adenocarcinoma patients; however, its roles and mechanisms in lung squamous cell carcinoma and large cell carcinoma need
Hidehito Yoshihara et al.
Biochemical and biophysical research communications, 423(1), 122-127 (2012-05-29)
Insulin receptor substrates (IRSs) play central roles in insulin/insulin-like growth factor (IGF) signaling and mediate a variety of their bioactivities. IRSs are tyrosine-phosphorylated by activated insulin receptor/IGF-I receptor tyrosine kinase in response to insulin/IGF, and are recognized by signaling molecules
Xin Li et al.
Asian Pacific journal of cancer prevention : APJCP, 13(5), 1749-1752 (2012-08-21)
Colorectal cancer (CRC), now the third most common cancer across the world, is known to aggregate in families. USP7 is a very important protein with an important role in regulating the p53 pathway, which is critical for genomic stability and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service