Skip to Content
Merck
All Photos(5)

Key Documents

HPA015597

Sigma-Aldrich

Anti-PLCE1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase epsilon-1, Anti-PLC-epsilon-1, Anti-Pancreas-enriched phospholipase C, Anti-Phosphoinositide-specific phospholipase C epsilon-1, Anti-Phospholipase C-epsilon-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

MGISPLGNQSVIIETGRAHPDSRRAVFHFHYEVDRRMSDTFCTLSENLILDDCGNCVPLPGGEEKQKKNYVAYTCKLMELAKNCDNKNEQLQCDHCDTLNDKYFCFEGSCEKVDMVYSGDSFCRKDFTDSQAAKT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLCE1(51196)

Immunogen

1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase epsilon-1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

1-Phosphatidylinositol-4,5-bisphosphate phosphodiesterase epsilon-1 is an enzyme encoded by the PLCE1 gene in humans and belongs to phospholipase family. This gene encodes a novel ras-related protein effector that helps in mediating the effects of R-Ras on the actin cytoskeleton as well as on membrane protrusion. It plays a crucial role in carcinogenesis and progression of several cancers (such as esophageal cancer, gastric cancer). It acts as an oncogene and regulates proteins which controls cell cycling and apoptosis, thus leading to cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72657

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen-Xing Liang et al.
Molecular cancer, 20(1), 103-103 (2021-08-21)
Constitutive activation of nuclear factor-κB (NF-κB) signaling plays a key role in the development and progression of colorectal carcinoma (CRC). However, the underlying mechanisms of excessive activation of NF-κB signaling remain largely unknown. We used high throughput RNA sequencing to
Masahiro Wakita et al.
The Journal of biological chemistry, 291(24), 12586-12600 (2016-04-08)
Phospholipase Cϵ (PLCϵ), an effector of Ras and Rap small GTPases, plays a crucial role in inflammation by augmenting proinflammatory cytokine expression. This proinflammatory function of PLCϵ is implicated in its facilitative role in tumor promotion and progression during skin
Manzoor Ahmad Malik et al.
Asian Pacific journal of cancer prevention : APJCP, 15(10), 4319-4323 (2014-06-18)
Phospholipase C epsilon 1 (PLCE1) encodes a member of the phospholipase family of proteins that play crucial roles in carcinogenesis and progression of several cancers including esophageal cancer (EC). In two large scale genome-wide association studies (GWAS) single nucleotide polymorphisms
Xinyang Liu et al.
Chinese medical journal, 127(13), 2511-2517 (2014-07-06)
The pathogenesis of gastric cancer (GC) involves environmental and genetic factors. Recently, two genome-wide association studies found that phospholipase C epsilon 1 (PLCE1) polymorphisms might be related to GC risk, and several studies further validated this finding. However, these studies
Li Zhao et al.
Asian Pacific journal of cancer prevention : APJCP, 15(13), 5437-5442 (2014-07-22)
Esophageal squamous cell carcinoma (ESCC) is one of the most malignancies with a poor prognosis. The phospholipase C? gene (PLCE1) encodes a novel ras-related protein effector mediating the effects of R-Ras on the actin cytoskeleton and membrane protrusion. However, molecular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service