Skip to Content
Merck
All Photos(4)

Key Documents

HPA014858

Sigma-Aldrich

Anti-BCAR3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Breast cancer anti-estrogen resistance protein 3, Anti-Novel SH2-containing protein 2, Anti-SH2 domain- containing protein 3B

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

SPLAEHRPDAYQDVSIHGTLPRKKKGPPPIRSCDDFSHMGTLPHSKSPRQNSPVTQDGIQESPWQDRHGETFTFRDPHLLDPTVEYVKFSKERHIMDRTPEKLKKEL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BCAR3(8412)

General description

BCAR3 (breast cancer anti-estrogen resistance 3) belongs to the SH2 domain-containing protein (NSP) family. In humans, this family contains three members namely, NSP1, NSP2/BCAR3 and NSP3. BCAR3 shows high homology to Cdc25 family of Rac GEFs, and contains a GEF (GDP-exchange factor)-like domain in its C-terminal. It also contains an SH2 domain, and a proline/serine-rich domain. It was first described in two estrogen-dependent human breast cancer cell lines, as the gene responsible for anti-estrogen resistance. It has a molecular weight of 95kDa.

Immunogen

Breast cancer anti-estrogen resistance protein 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BCAR3 (breast cancer anti-estrogen resistance 3) is up-regulated in breast cancers, and is related to invasive and advanced breast cancer type. It facilitates advancement of breast cancer, by activating intracellular signaling pathways essential for metastasis. It is an interacting partner of p130Cas, which in turn activates Rac1. It regulates cell survival, proliferation and movement, by facilitating the interaction between Cas and the protein tyrosine kinase c-Src. It facilitates the progression of cell cycle by interacting with epidermal growth factor (EGF) through its SH2 domain. It promotes cell division in normal human breast MCF-12A cells. It is responsible for anti-estrogen resistance in breast cancer cells by activating PI3K (phosphatidylinositol 3-kinase), which in turn activates Rac pathway.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72807

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashley L Wilson et al.
PloS one, 8(6), e65678-e65678 (2013-06-14)
Metastatic breast cancer is incurable. In order to improve patient survival, it is critical to develop a better understanding of the molecular mechanisms that regulate metastasis and the underlying process of cell motility. Here, we focus on the role of
Myung-Ju Oh et al.
Biochemical and biophysical research communications, 375(3), 430-434 (2008-08-30)
BCAR3 (breast cancer anti-estrogen resistance 3) is a signal transducer containing an SH2 domain, a proline/serine-rich domain and a GDP-exchange factor homologous domain, whose role in signaling pathways is currently unclear. Furthermore, BCAR3 is implicated in anti-estrogen resistance of breast
P Pennekamp et al.
Sexual development : genetics, molecular biology, evolution, endocrinology, embryology, and pathology of sex determination and differentiation, 5(4), 197-204 (2011-06-10)
We identified Bcar3 in the course of a screen for developmentally regulated genes at early developmental stages in mouse embryos. In this study, we explored the spatio-temporal expression pattern of Bcar3 during the critical time period of sex determination using
Anthony Makkinje et al.
Cellular signalling, 21(9), 1423-1435 (2009-05-21)
NSP protein family members associate with p130Cas, a focal adhesion adapter protein best known as a Src substrate that integrates adhesion-related signaling. Over-expression of AND-34/BCAR3/NSP2 (BCAR3), but not NSP1 or NSP3, induces anti-estrogen resistance in human breast cancer cell lines.
Kyriacos N Felekkis et al.
Molecular cancer research : MCR, 3(1), 32-41 (2005-01-27)
AND-34, a 95-kDa protein with modest homology to Ras GDP exchange factors, associates with the focal adhesion protein p130Cas. Overexpression of AND-34 confers anti-estrogen resistance in breast cancer cell lines, a property linked to its ability to activate Rac. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service