Skip to Content
Merck
All Photos(5)

Key Documents

HPA014467

Sigma-Aldrich

Anti-KLHDC8B antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Kelch domain-containing protein 8B

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on16 April 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Estimated to ship on16 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

MPTPCYGASTFLHGNKIYVLGGRQGKLPVTAFEAFDLEARTWTRHPSLPSRRAFAGCAMAEGSVFSLGGLQQPGPHNFYSRPHFVNTVEMFDLEHGSWTKLPRSLRMRDKRADFVVGSL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The gene KLHDC8B (kelch domain containing 8B) encodes a protein containing seven kelch repeat domains that is expressed during mitosis. The gene is mapped to human chromosome 3p21.31, a region implicated in lymphoma and related malignancies. It localizes to the midbody structure during cytokinesis.

Immunogen

Kelch domain-containing protein 8B recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene KLHDC8B (kelch domain containing 8B) encodes a protein that prevents chromosomal instability, centrosomal amplification and mitotic errors. Decrease in the expression of this protein leads to an increase in binucleated cells, suggesting its role in cytokinesis. This depletion produces binucleated, giant Reed-Sternberg (RS) cell, associated with Hodgkin lymphoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71366

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen J Salipante et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(35), 14920-14925 (2009-08-27)
Classical Hodgkin lymphoma (cHL) is a malignancy of B-cell origin in which the neoplastic cells, known as "Reed-Sternberg" (RS) cells, are characteristically binucleated. Here we describe a family where multiple individuals developing cHL have inherited a reciprocal translocation between chromosomes
Andrew E Timms et al.
Communicative & integrative biology, 3(2), 154-158 (2010-06-30)
A key feature of Hodgkin lymphoma is that the malignant cells are binucleated, as a consequence of failed cytokinesis. We recently ascertained a family in which multiple cases of Hodgkin lymphoma had occurred among individuals who inherited a balanced chromosomal
Maxwell M Krem et al.
The Journal of biological chemistry, 287(46), 39083-39093 (2012-09-19)
The malignant cell in classical Hodgkin lymphoma (HL) is the binucleated giant Reed-Sternberg cell. Chromosomal instability and mitotic errors may contribute to HL pathogenesis; one potential mitotic regulator is the kelch protein KLHDC8B, which localizes to the midbody, is expressed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service