Skip to Content
Merck
All Photos(6)

Key Documents

HPA014319

Sigma-Aldrich

Anti-MEGF9 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-EGF-like domain-containing protein 5, Anti-Multiple EGF-like domain protein 5, Anti-Multiple epidermal growth factor-like domains 9 precursor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

YQNRKLNAPFWTIELKEDNISFSSYHDSIPNADVSGLLEDDGNEVAPNGQLTLT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MEGF9(1955)

General description

The gene MEGF9 (multiple epidermal growth factor-like domains 9) encodes a transmembrane protein containing multiple EGF-like repeats. The N-terminus contains several potential O-glycosylation sites followed by five EGF-like domains. It also contains a single pass transmembrane domain followed by a highly conserved short intracellular domain that has potential phosphorylation sites. The mRNA is found to be expressed in the developing and adult CNS (central nervous system) and PNS (peripheral nervous system), particularly in Purkinje cells of the cerebellum and in glial cells of the PNS. It is also found to some extent in the epidermal layer of skin, papillae of the tongue and the epithelium of the gastrointestinal tract. The gene is mapped to human chromosome 9q32–9q33.3. The human gene consists of six exons spanning a length of 113.66kb. The signal peptide sequence and an N-terminal domain are encoded by exon 1, exons 2–5 encode EGF-like repeats 1–5, and exon 6 encodes a transmembrane region and a cytoplasmic tail. A long untranslated region of 4326bp is also encoded by exon 6. The 3′ end contains a conserved polyadenylation site.

Immunogen

Multiple epidermal growth factor-like domains 9 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene MEGF9 (multiple epidermal growth factor-like domains 9) encodes a novel putative receptor that may function as a guidance or signaling molecule. It is regulated during development. The exact function of this protein is yet to be characterized. Proteins containing EGF-like domains usually participate in the development, maintenance and injury response of the nervous system.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72839

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ulrike Brandt-Bohne et al.
The Biochemical journal, 401(2), 447-457 (2006-09-20)
MEGF9 [multiple EGF (epidermal growth factor)-like-domains 9], a novel transmembrane protein with multiple EGF-like repeats, is predominantly expressed in the developing and adult CNS (central nervous system) and PNS (peripheral nervous system). The domain structure of MEGF9 consists of an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service