Skip to Content
Merck
All Photos(1)

Key Documents

HPA013602

Sigma-Aldrich

Anti-DAT antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DAT1, Anti-SLC6A3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

LHYLFSSFTTELPWIHCNNSWNSPNCSDAHPGDSSGDSSGLNDTFGTTPAAEYFERGVLHLHQSHGIDDLG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SLC6A3(6531)

General description

The dopamine transporter (DAT) gene is mapped to human chromosome 5p15 and the untranslated region at the 3′ end contains a 40 bp tandem repeat that is referred to as a variable number tandem repeat or VNTR.

Immunogen

Sodium-dependent dopamine transporter recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene DAT is also called as SLC6A3 (solute carrier family 6 (neurotransmitter transporter), member 3) and functions as a dopamine transporter. It belongs to the sodium- and chloride-dependent neurotransmitter transporter family. It functions in the transport of released dopamine back up into presynaptic terminals. The VNTR (variable number tandem repeat) copy number in the gene may vary and this variation has been linked to various disorders including anxiety, social phobia, obsessive-compulsive, and Tourette′s. It is also associated with Parkinson′s disease and related neurodegenerative disorders.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72519.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J Vandenbergh et al.
Genomics, 14(4), 1104-1106 (1992-12-01)
The human dopamine transporter (DAT1) gene is localized to chromosome 5p15.3 by in situ hybridization and PCR amplification of rodent somatic cell hybrid DNA. Analysis of a 40-bp repeat in the 3' untranslated region of the message revealed variable numbers
D J Nicholl et al.
Neurology, 53(7), 1415-1421 (1999-10-26)
To determine whether reported genetic association of polymorphisms in the CYP2D6, CYP1A1, N-acetyltransferase 2 (NAT2), DAT1, and glutathione s-transferase M1 (GSTM1) genes with PD were evident in a population of 176 unrelated patients with sporadic PD and to extend these
D C Rowe et al.
Behavior genetics, 28(3), 215-225 (1998-07-22)
The relation of the dopamine transporter gene (DAT1) to symptoms of internalizing disorders, Tourette's disorder, and obsessive-compulsive disorder was examined using both within- and between-family tests of association. The sample consisted of clinic-referred children and their siblings and controls and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service