Skip to Content
Merck
All Photos(2)

Key Documents

HPA012948

Sigma-Aldrich

Anti-TNFSF15 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-TNF ligand-related molecule 1, Anti-Tumor necrosis factor ligand superfamily member 15, Anti-Vascular endothelial cell growth inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

FRGMTSECSEIRQAGRPNKPDSITVVITKVTDSYPEPTQLLMGTKSVCEVGSNWFQPIYLGAMFSLQEGDKLMVNVSDISLVDYTKEDKTFFGAFLL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNFSF15(9966)

General description

Tumor necrosis factor ligand superfamily member 15 (TNFSF15) is a 174 amino acid, type II transmembrane protein. The molecular weight of the protein is 13kDa and it belongs to the tumor necrosis factor (TNF) family. TNFSF15 has a carboxyl terminal which is present on the exterior of the cell surface, a transmembrane domain and a short cytoplasmic tail. The gene encoding it is located on chromosome 9q32.

Immunogen

Tumor necrosis factor ligand superfamily member 15 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Tumor necrosis factor ligand superfamily member 15 (TNFSF15) functions as an angiogenesis inhibitor by inhibiting the endothelial cell proliferation. TNFSF15 activates the transcription factor κB (nuclear factor-κβ). It induces the degradation of the inhibitory subunit IκΒα and helps in the nuclear translocation of the p65 subunit of NF-κβ. TNFSF15 also activates c-Jun N-terminal kinase and inhibits the growth of various human tumor cell. Single nucleotide polymorphisms in TNFSF15 are associated with surgical diverticulitis and susceptibility to ulcerative colitis and Crohn′s disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71528

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jixiang Zhang et al.
Autoimmunity, 47(8), 512-518 (2014-07-17)
Several polymorphisms have been identified in TNFSF15, while their roles in the incidence of ulcerative colitis (UC) and Crohn's disease (CD) are conflicting. This meta-analysis was aimed to clarify the impact of these polymorphisms on UC and CD risk. Databases
V Haridas et al.
Oncogene, 18(47), 6496-6504 (1999-12-22)
Recently a new member of the human tumor necrosis factor (TNF) family named as VEGI was reported. However, very little is known about the biological activities displayed by this cytokine. In this report, we show that in myeloid cells VEGI
Y Zhai et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 13(1), 181-189 (1999-01-05)
A novel member of the tumor necrosis factor (TNF) family has been identified from the human umbilical vein endothelial cell cDNA library, named vascular endothelial growth inhibitor (VEGI). The VEGI gene was mapped to human chromosome 9q32. The cDNA for
Tara M Connelly et al.
Annals of surgery, 259(6), 1132-1137 (2014-05-13)
To determine if single nuclear polymorphisms (SNPs) in the TFNSF15 gene play a role in patients requiring surgery for diverticulitis. A role for a genetic predisposition in diverticulitis is suggested by its association with hereditary connective tissue disorders, youthful onset

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service