Skip to Content
Merck
All Photos(4)

Key Documents

HPA012743

Sigma-Aldrich

Anti-SCNN1A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Alpha-ENaC, Anti-Alpha-NaCH, Anti-Amiloride-sensitive sodium channel subunit alpha, Anti-Epithelial Na(+) channel subunit alpha, Anti-Nonvoltage-gated sodium channel 1 subunit alpha, Anti-SCNEA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Check Cart for Availability


Select a Size

Change View
100 μL
$952.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$952.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

RYPEIKEELEELDRITEQTLFDLYKYSSFTTLVAGSRSRRDLRGTLPHPLQRLRVPPPPHGARRARSVASSLRDNNPQVDWKDWKIGFQL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCNN1A(6337)

General description

SCNN1A is an amiloride-sensitive Na+ channel that regulates the translocation of electrolytes and fluids across the epithelium. SCNN1A is expressed in human pulmonary, pancreatic, hepatic and renal tissues . This channel protein is also involved in the synthesis of ion conductive pores across membranes . Anti-SCNN1A antibody is specific for SCCN1A in humans.

Immunogen

Amiloride-sensitive sodium channel subunit alpha recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71790

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng Xu et al.
Biomedicines, 10(5) (2022-05-29)
Salt sensitivity of blood pressure (BP) refers to an increase in BP following an increase in dietary salt, which is associated with increased incidence of cardiovascular disease and early death. However, decreased sodium intake also increases mortality and morbidity. Inverse
Jessica Welzel et al.
International journal of cosmetic science, 44(2), 216-231 (2022-03-10)
Human eccrine sweat glands (eSG) represent vital components of the skin involved in regulating body temperature. Especially the eccrine duct, which opens directly into the skin surface and releases the aqueous sweat, constitutes the first contact point with topically applied
J K Tucker et al.
The American journal of physiology, 274(4 Pt 1), C1081-C1089 (1998-05-12)
The alpha-subunit of the amiloride-sensitive epithelial Na+ channel (alpha ENaC) is critical in forming an ion conductive pore in the membrane. We have identified the wild-type and three splice variants of the human alpha ENaC (h alpha ENaC) from the
F J McDonald et al.
The American journal of physiology, 266(6 Pt 1), L728-L734 (1994-06-11)
Amiloride-sensitive Na+ channels control, in part, fluid and electrolyte transport across epithelia in many organs. In the lung, they control the quantity and composition of the respiratory tract fluid and play a key role in the transition from a fluid-filled
Ilaria Musante et al.
Experimental physiology, 104(6), 866-875 (2019-03-30)
What is the central question of this study? What is the precise subcellular localization of the epithelial sodium channel (ENaC) in human airway epithelium? What is the main finding and its importance? ENaC protein has an unexpected localization in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service