Skip to Content
Merck
All Photos(5)

Key Documents

HPA012612

Sigma-Aldrich

Anti-CADM4 antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cell adhesion molecule 4 precursor, Anti-Immunoglobulin superfamily member 4C, Anti-Nectin-like protein 4, Anti-TSLC1-like protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on30 April 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Estimated to ship on30 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

GGIIICEAQNQALPSGHSKQTQYVLDVQYSPTARIHASQAVVREGDTLVLTCAVTGNPRPNQIRWNRGNESLPERAEAVGETLTLPGLVSADNGTYTCEASNKHGHARALYVLVVYDPGAVVEAQTSVPY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CADM4(199731)

General description

Cell adhesion molecule 4 precursor (CADM4) is a 55kDa protein expressed in the kidneys, bladder, brain and prostate gland. It belongs to the tumor suppressor in lung cancer 1 (TSLC1) gene family and the gene encoding it is present on chromosome 19q13.2. CADM4 has three Ig (immunoglobulin) loops in the extracellular domain, one transmembrane domain and a short cytoplasmic domain.

Immunogen

Cell adhesion molecule 4 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Cell adhesion molecule 4 precursor (CADM4) interacts by the formation of homo-dimers and its overexpression induces the aggregation of cells in a Ca2+/Mg2+-independent manner. It is thus involved in cell adhesion through homophilic trans-interaction. CADM4 suppresses tumor growth and in human proximal uriniferous tubules, its cytoplasmic region interacts with 4.1B, an actin binding protein which links CADM4 to different pathways. It mediates the contacts between Schwann cells and axons. The myelin unit establishment in the peripheral nervous system is taken care by CADM4. CADM4 interacts in cis with Erb-b2 receptor tyrosine kinase 3 (ErbB3), recruits protein tyrosine phosphatase non-receptor type 13 (PTPN13) and inhibits the heregulin-induced activation of the ErbB2/ErbB3 signaling pathway. It also interacts in cis with integrin-α6β4 and inhibits the disassembly of hemi desmosomes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71553

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neev Golan et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(27), 10950-10961 (2013-07-05)
The interaction between myelinating Schwann cells and the axons they ensheath is mediated by cell adhesion molecules of the Cadm/Necl/SynCAM family. This family consists of four members: Cadm4/Necl4 and Cadm1/Necl2 are found in both glia and axons, whereas Cadm2/Necl3 and
Hirokazu Sugiyama et al.
Genes to cells : devoted to molecular & cellular mechanisms, 18(6), 519-528 (2013-04-25)
Nectin-like molecule 4 (Necl-4)/CADM4, a transmembrane cell-cell adhesion molecule with three Ig-like domains, was shown to serve as a tumor suppressor, but its mode of action has not been elucidated. In this study, we showed that Necl-4 interacted in cis
Y N Williams et al.
Oncogene, 25(10), 1446-1453 (2005-11-02)
The TSLL2/IGSF4C encodes an immunoglobulin (Ig) superfamily molecule showing significant homology with a lung tumor suppressor, TSLC1. The TSLL2 protein of 55 kDa is mainly expressed in the kidney, bladder, and prostate in addition to the brain. Here, we report
Masayoshi Nagata et al.
International journal of cancer, 130(6), 1329-1337 (2011-05-06)
Renal clear cell carcinoma (RCCC) is the most frequent subpopulation of renal cell carcinoma and is derived from the proximal uriniferous tubules. We have previously reported that an actin-binding protein, 4.1B/DAL-1, is expressed in renal proximal tubules, whereas it is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service