Skip to Content
Merck
All Photos(11)

Key Documents

HPA012609

Sigma-Aldrich

Anti-TGOLN2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-TGN38 homolog, Anti-TGN46, Anti-TGN48, Anti-Trans-Golgi network integral membrane protein 2 precursor, Anti-Trans-Golgi network protein TGN51

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

ATESVKQEEAGVRPSAGNVSTHPSLSQRPGGSTKSHPEPQTPKDSPSKSSAEAQTPEDTPNKSGAEAKTQKDSSNRSGA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TGOLN2(10618)

General description

Trans-Golgi network integral membrane protein 2 (TGOLN2) is a heterodimeric type I integral membrane protein. It has a highly conserved N terminus which consists of a signal peptide. The C terminus comprises part of the lumenal domain, a membrane spanning region and cytoplasmic tail.

Immunogen

Trans-Golgi network integral membrane protein 2 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-TGOLN2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Trans-Golgi network integral membrane protein 2 (TGOLN2) cycles between the trans-golgi network (TGN) and the cell surface via an early endosomal compartment. This movement is mediated by a tyrosine-based tetra peptide signal (SDYQRL) in the cytoplasmic domain. It plays an important role in the formation of exocytic vesicles at the TGN by functioning as a receptor for complexes of a cytoplasmic protein known as p62 and a GTP-binding protein. The cytoplasmic domain of TGOLN2 binds to the complex and is essential for budding to occur. Coupling of the segregation of secretory proteins to the budding of exocytic vesicles is mediated by it. The cytosolic domain of TGOLN2 interacts with a complex of four adaptins, called AP2 clathrin adaptor complex and also with the coiled coil region of a protein called neurabin.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72106

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monique Philippe et al.
The Journal of biological chemistry, 288(18), 13046-13056 (2013-03-28)
Weibel-Palade bodies (WPBs) are specific cigar-shaped granules that store von Willebrand factor (VWF) for its regulated secretion by endothelial cells. The first steps of the formation of these granules at the trans-Golgi network specifically require VWF aggregation and an external
S Ponnambalam et al.
Journal of cell science, 109 ( Pt 3), 675-685 (1996-03-01)
cDNAs encoding the human and macaque homologues of rat TGN38 have been cloned and sequenced. The proteins have a highly conserved N terminus (comprising the signal peptide) and C terminus (comprising part of the lumenal domain, the membrane spanning region
D J Stephens et al.
The Journal of biological chemistry, 274(42), 30080-30086 (1999-10-09)
TGN38 is a type I integral membrane protein that constitutively cycles between the trans-Golgi network (TGN) and plasma membrane. The cytosolic domain of TGN38 interacts with AP2 clathrin adaptor complexes via the tyrosine-containing motif (-SDYQRL-) to direct internalization from the
K K Stanley et al.
Trends in cell biology, 3(8), 252-255 (1993-08-01)
TGN38/41 is a heterodimeric integral membrane protein that cycles between the trans Golgi network and the cell surface. A tyrosine-containing tetrapeptide motif within its cytoplasmic tail is necessary and sufficient for determining its steady-state location in the TGN. Recent results

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service