Skip to Content
Merck
All Photos(7)

Documents

HPA011990

Sigma-Aldrich

Anti-ACADL antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LCAD, Anti-Long-chain specific acyl-CoA dehydrogenase, mitochondrial precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GEERLETPSAKKLTDIGIRRIFSPEHDIFRKSVRKFFQEEVIPHHSEWEKAGEVSREVWEKAGKQGLLGVNIAEHLGGIGGDLYSAAIVWEEQAYSNCSGPGFSIHSGIVMSYITNHGSEEQIKHFIPQMT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACADL(33)

Immunogen

Long-chain specific acyl-CoA dehydrogenase, mitochondrial precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ACADL (Acyl-CoA dehydrogenase, long chain) is a mitochondrial flavoenzyme. It is highly expressed in human skeletal muscles and to a lesser extent in liver, fibroblast and heart tissues. It is involved in the first stage of branched long-chain fatty acid (LCFA) β-oxidation. During fatty acid β-oxidation, it interacts with 2,6-dimethylheptanoyl-CoA and catalyzes the degradation of double-bonded unsaturated fatty acid chain in the mitochondria. Mutation in ACADL causes deficiency of VLCAD (very long chain acyl-CoA dehydrogenase) and MCAD (Medium chain acyl-CoA dehydrogenase) with disrupted muscle function, myopathy, and cardiomyopathy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70470

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kegong Chen et al.
Cancer cell international, 24(1), 86-86 (2024-02-25)
The role of Acyl-CoA dehydrogenase long chain (ACADL) in different tumor types had different inhibiting or promoting effect. However, its role in non-small cell lung cancer (NSCLC) carcinogenicity is not clear. In this study, we utilized The Cancer Genome Atlas
Xiaofang Zhao et al.
NPJ precision oncology, 4, 7-7 (2020-03-29)
Long-chain acyl-CoA dehydrogenase (ACADL) is a mitochondrial enzyme that catalyzes the initial step of fatty acid oxidation, but the role of ACADL in tumor biology remains largely unknown. Here, we found that ACADL was frequently downregulated in hepatocellular carcinoma (HCC)
W Lea et al.
Biochimica et biophysica acta, 1485(2-3), 121-128 (2000-06-01)
The first reaction of mitochondrial beta-oxidation, which is catalyzed by acyl-CoA dehydrogenases, was studied with unsaturated fatty acids that have a double bond either at the 4,5 or 5,6 position. The CoA thioesters of docosahexaenoic acid, arachidonic acid, 4,7,10-cis-hexadecatrienoic acid
Amy C Maher et al.
Molecular genetics and metabolism, 100(2), 163-167 (2010-04-07)
Long-chain acyl-CoA dehydrogenase (LCAD) is a mitochondrial flavoenzyme thought to be one of the major enzymes responsible for the first step of long-chain fatty acid (LCFA) beta-oxidation. Surprisingly, recent studies have shown LCAD is hardly detectable in human tissues such
Validating MCM2 as a clinically relevant surrogate immunohistochemical marker for an aggressive meningioma molecular subtype.
Allison Shelbourn et al.
Journal of neuropathology and experimental neurology, 82(12), 1037-1039 (2023-10-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service