Skip to Content
Merck
All Photos(2)

Key Documents

HPA011781

Sigma-Aldrich

Anti-FAM110B antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Protein FAM110B

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

KYVKSQEVINAKQEPVKPAVLAKPPVCPAAKRALGSPTLKVFGNHAKTESGVQRENLKLEILKNIINSSEGSSSGSGHKHSSRNWPPHRSEATDLHRHSFAESLKVYPTQGRRSPQEGGSHVGRRLLEQSAESFLHVSHSSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FAM110B(90362)

Immunogen

Protein FAM110B recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FAM110B (Family with sequence similarity 110, member B) encodes a protein with distinct conserved motifs. It is expressed in the centrosomes and spindle poles. It forms clusters at the microtubule organization center in interphase and at spindle poles in mitosis and thus, restricting the cell cycle progression in G1 phase. It is an essential factor in the progression of castration-resistant prostate cancer (CRPC). The activity of FAM110B is regulated by androgens and in turn, it controls the androgen signal responses in prostate cancer cells. Studies show the therapeutic aspects of FAM110B in multiple cancers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71355

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paula Vainio et al.
The Prostate, 72(7), 789-802 (2011-09-16)
Castration-resistant prostate cancer (CRPC) represents a therapeutic challenge for current medications. In order to explore the molecular mechanisms involved in CRPC progression and to identify new therapeutic targets, we analyzed a unique sample set of 11 CRPCs and 7 advanced
Helena Hauge et al.
Genomics, 90(1), 14-27 (2007-05-15)
We have previously characterized the centrosome/spindle pole-associated protein (CSPP) involved in cell cycle progression. The open reading frame C20orf55 was identified in a yeast two-hybrid screen in a search for CSPP-interacting proteins. A homology search revealed that C20orf55 belongs to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service