Skip to Content
Merck
All Photos(9)

Key Documents

HPA011276

Sigma-Aldrich

Anti-SSR1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-SSR-alpha, Anti-Signal sequence receptor subunit alpha, Anti-TRAP-alpha, Anti-Translocon-associated protein subunit alpha precursor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Estimated to ship on30 May 2025



Select a Size

Change View
100 μL
$952.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$952.00


Estimated to ship on30 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

ESRKRKRPIQKVEMGTSSQNDVDMSWIPQETLNQINKASPRRLPRKRAQKRSVGSDE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SSR1(6745)

General description

SSR1 (signal sequence receptor, α) is an endoplasmic reticulum (ER) membrane protein, which is predominantly localized to rough ER. Its molecular weight is 34kDa. It is a type I membrane protein and is also found in the nuclear envelope. This gene is localized to human chromosome 6. It is also known as translocon-associated protein α (TRAPA) and is one of the four subunits making the TRAP complex.

Immunogen

Translocon-associated protein subunit alpha precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-SSR1 antibody produced in rabbit has been used in selective permeabilization immunofluorescence (2μg/ml).
Anti-SSR1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

The function of SSR1 (signal sequence receptor, α) is not yet fully characterized. It is a calcium binding protein. It forms part of the translocon-associated protein (TRAP) complex, which is involved in the endoplasmic reticulum-associated degradation (ERAD) pathway. This protein is induced by granulocyte-macrophage colony-stimulating factor (GM-CSF), which might be an outcome of unfolded protein response due to the accumulation of improperly folded proteins. Also this gene is located in the schizophrenia susceptibility locus.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71885

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koji Nagasawa et al.
EMBO reports, 8(5), 483-489 (2007-03-24)
The mammalian translocon-associated protein (TRAP) complex comprises four transmembrane protein subunits in the endoplasmic reticulum. The complex associates with the Sec61 translocon, although its function in vivo remains unknown. Here, we show the involvement of the TRAP complex in endoplasmic
Martin S Taylor et al.
The Journal of biological chemistry, 288(45), 32211-32228 (2013-09-21)
Ghrelin O-acyltransferase (GOAT) is a polytopic integral membrane protein required for activation of ghrelin, a secreted metabolism-regulating peptide hormone. Although GOAT is a potential therapeutic target for the treatment of obesity and diabetes and plays a key role in other
F Vogel et al.
European journal of cell biology, 53(2), 197-202 (1990-12-01)
The signal sequence receptor (SSR), an integral membrane glycoprotein of 34 kDa, has previously been shown to be a component of the molecular environment which nascent polypeptide chains meet in passage through the endoplasmic reticulum (ER) membrane. We have used
E Hartmann et al.
FEBS letters, 349(3), 324-326 (1994-08-08)
The alpha-subunit of the TRAP complex (TRAP alpha) is a single-spanning membrane protein of the endoplasmic reticulum (ER) which is found in proximity of nascent polypeptide chains translocating across the membrane. Here, we demonstrate the widespread occurrence of TRAP alpha
T Hirama et al.
FEBS letters, 455(3), 223-227 (1999-08-07)
The cloning of full length cDNA for the translocon-associated protein alpha subunit, previously called signal sequence receptor alpha, is reported as a result of differential display experiments in search of genes induced by granulocyte-macrophage colony-stimulating factor. Its messenger RNA was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service