Skip to Content
Merck
All Photos(3)

Key Documents

HPA011273

Sigma-Aldrich

Anti-HSPA12A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Heat shock 70 kDa protein 12A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on31 May 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Estimated to ship on31 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

KLLVKDGTRWCTDVFDKFISADQSVALGELVKRSYTPAKPSQLVIVINIYSSEHDNVSFITDPGVKKCGTLRLDLTGTSGTAVPARREIQTLMQFGDTEIKATAIDIATSKSVKVGID

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

HSPA12A (heat shock 70kDa protein 12A), along with HSPA12B, forms a subfamily of Hsp70 protein family. It is expressed in brain, heart, kidney and muscle.

Immunogen

Heat shock 70 kDa protein 12A recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

HSPA12A (heat shock 70kDa protein 12A) is highly expressed in the frontal and occipital regions of brain cortex. However, it is down-regulated in prefrontal cortex of schizophrenic subjects. Its expression is elevated in atherosclerotic lesions, and it might play a role in the susceptibility to atherosclerosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71592

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie L Pongrac et al.
Biological psychiatry, 56(12), 943-950 (2004-12-17)
Deoxyribonucleic acid microarray analyses of dorsolateral prefrontal cortex (DLPFC) area 9 from 10 matched pairs of schizophrenic and control subjects revealed a consistent and significant decrease (p = .001; mean log2 signal difference = -.58) in transcript expression for a
Zhihua Han et al.
Proceedings of the National Academy of Sciences of the United States of America, 100(3), 1256-1261 (2003-01-29)
Gene expression profiling was carried out comparing Con A elicited peritoneal macrophages from C57BL6 and FVBN wild-type and apolipoprotein (apo)E knockout mice. An EST, was expressed at higher levels in C57BL6 compared with FVBN mice. mapped to an atherosclerosis susceptibility
Peder Madsen et al.
Scientific reports, 9(1), 611-611 (2019-01-27)
SorLA and Sortilin are multifunctional receptors involved in endocytosis and intracellular sorting of different and unrelated ligands. SorLA has recently attracted much attention as a novel strong risk gene for Alzheimer's disease, and much effort is currently being put into

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service