Skip to Content
Merck
All Photos(6)

Key Documents

HPA011157

Sigma-Aldrich

Anti-LAT antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-36 kDa phospho-tyrosine adapter protein antibody produced in rabbit, Anti-Linker for activation of T-cells family member 1 antibody produced in rabbit, Anti-p36-38 antibody produced in rabbit, Anti-pp36 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Check Cart for Availability


Select a Size

Change View
100 μL
$952.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$952.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

HCHRLPGSYDSTSSDSLYPRGIQFKRPHTVAPWPPAYPPVTSYPPLSQPDLLPIPRSPQPLGGSHRTPSSRRDSDG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SPNS1(83985)

General description

LAT (linker for activation of T cells) is a leukocyte type III transmembrane adaptor protein. It is made of 262 amino acids, and has a shorter isoform made of 233 amino acids. It is a lipid raft associated protein, and does so by its two cysteine residues present proximal to the membrane. This protein resides in the plasma membrane as well as intracellular and subsynaptic vesicles. It has a small exoplasmic region, a single membrane spanning domain, and a cytoplasmic region containing nine tyrosine residues. This gene maps to human chromosome 16p11.2, spans 5.7kb and has 11 exons.

Immunogen

Linker for activation of T-cells family member 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

LAT (linker for activation of T cells) plays an essential role in the development and activation of T-cells, and T-cell receptor (TCR) signaling. Phosphorylated Zap-70 (Zeta-chain-associated protein kinase 70) phosphorylates the tyrosine residues of LAT. This activated LAT then initiates TCR-signaling. Activated LAT induces multiple signaling intermediates, leading to Ca2+ influx, activation of NFAT (nuclear factor of activated T cells), which eventually leads to the activation of cytokines. It regulates the proliferation of lymphocytes, and controls the differentiation of T-cells into T-helper 1 (Th1) and Th2 subtypes. Activation of natural killer (NK) cells, lead to the phosphorylation and activation of LAT, which in turn interacts with various phosphotyrosine containing proteins, such as PCLγ (phospholipase Cγ). This leads to an overall increase in the cytotoxic function of NK cells. LAT is overexpressed in severe aplastic anemia (SAA), where up-regulated LAT might lead to increased activity of T-cells, and disruption of Th1/Th2 balance.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72225

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Václav Horejsí et al.
The FEBS journal, 277(21), 4383-4397 (2010-09-30)
Membrane rafts are microdomains involved in a number of biologically important processes, including immunoreceptor signalling. Among the functionally important protein components of these microdomains are transmembrane adaptor proteins, containing in their intracellular domains tyrosine residues that can be phosphorylated and
David J Williamson et al.
Nature immunology, 12(7), 655-662 (2011-06-07)
Engaged T cell antigen receptors (TCRs) initiate signaling through the adaptor protein Lat. In quiescent T cells, Lat is segregated into clusters on the cell surface, which raises the question of how TCR triggering initiates signaling. Using super-resolution fluorescence microscopy
Lakshmi Balagopalan et al.
Journal of immunology (Baltimore, Md. : 1950), 190(8), 3849-3853 (2013-03-15)
A controversy has recently emerged regarding the location of the cellular pool of the adapter linker for activation of T cells (LAT) that participates in propagation of signals downstream of the TCR. In one model phosphorylation and direct recruitment of
C Windpassinger et al.
Cytogenetic and genome research, 97(3-4), 155-157 (2002-11-20)
We have mapped the LAT gene by radiation hybrid mapping and fluorescence in situ hybridization to chromosome 16p11.2. The complete cDNA sequence of LAT was generated using assembled sequences of cDNA fragments already available. BLAST analysis using the cDNA sequence
Naoki Kunii et al.
Human gene therapy, 24(1), 27-37 (2012-09-25)
It is likely that the enhancement of signaling after antigenic stimulation, particularly in the tumor microenvironment, would improve the function of adoptively transferred T cells. Linker for activation of T cells (LAT) plays a central role in T cell activation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service