Skip to Content
Merck
All Photos(9)

Key Documents

HPA011036

Sigma-Aldrich

Anti-ARMC10 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-SVH protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on29 May 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Estimated to ship on29 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LTDGSYDDVLNAEQLQKLLYLLESTEDPVIIERALITLGNNAAFSVNQAIIRELGGIPIVANKINHSNQSIKEKALNALNNLSVNVEN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARMC10(83787)

Looking for similar products? Visit Product Comparison Guide

General description

ARMC10 (armadillo repeat containing 10) belongs to armadillo family of proteins, and forms a subfamily with armadillo repeat containing X-linked (ARMCX)1-6, which contain an uncharacterized domain in their C-termini. It is a transmembrane protein and resides in endoplasmic reticulum (ER), vacuoles, Golgi bodies and mitochondria. This gene maps to human chromosome 7q22.1, and is ubiquitously expressed in human tissues. It is predominantly expressed in brain and in developing neurons, where it resides in mitochondria and nuclei.

Immunogen

SVH protein recombinant protein epitope signature tag (PrEST)

Application

Anti-ARMC10 antibody is suitable for immunocytochemistry. Anti-ARMC10 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Armadillo proteins regulate multiple functions such as tumorigenesis and embryogenesis, through their armadillo repeat. ARMC10 (armadillo repeat containing 10) is overexpressed in hepatocellular carcinomas. It interacts with kinesin/Miro/Trak2 complex to control mitochondrial dynamics and trafficking. It also plays a protective role against Aβ-induced toxicity, and its up-regulation inhibits Aβ-induced fragmentation of mitochondria. ARMC10-B isoform interacts with p53, and inhibits it, and thus, prevents apoptosis and promotes cell growth.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72237

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Serrat et al.
Cell death & disease, 5, e1163-e1163 (2014-04-12)
Mitochondrial function and dynamics are essential for neurotransmission, neural function and neuronal viability. Recently, we showed that the eutherian-specific Armcx gene cluster (Armcx1-6 genes), located in the X chromosome, encodes for a new family of proteins that localise to mitochondria
Xinyuan Zhou et al.
FEBS letters, 581(25), 4943-4948 (2007-10-02)
We previously reported that inhibition of SVH-B, a specific splicing variant of SVH, results in apoptotic cell death. In this study, we reveal that this apoptosis may be dependent on the presence of p53. Co-immunoprecipitation and GST pull-down assays have
Yusuke Kusama et al.
Experimental and therapeutic medicine, 1(2), 395-399 (2010-03-01)
The armadillo family of proteins has been implicated in embryogenesis and tumorigenesis. Armadillo repeat containing X-linked (ARMCX)1-6 and its most closely related protein, ARMC10, share an uncharacterized domain in their carboxyl-terminal region and thereby constitute a unique subfamily. We previously

Articles

Learn about glioma markers for high-grade gliomas and low-grade gliomas and find reliable Prestige Antibodies® to target glioma markers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service