Skip to Content
Merck
All Photos(3)

Key Documents

HPA010773

Sigma-Aldrich

Anti-LRIG3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LIG-3, Anti-Leucine-rich repeats and immunoglobulin-like domains protein 3 precursor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

NLCLNKSSLDFSANPEPASVASSNSFMGTFGKALRRPHLDAYSSFGQPSDCQPRAFYLKAHSSPDLDSGSEEDGKERTDFQEENHICTFKQTLENYRTPN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LRIG3(121227)

General description

LRIG3 (leucine-rich repeats and immunoglobulin-like domains 3) belongs to a family of integral plasma membrane proteins called LRIG. It is a transmembrane glycol-protein, which consists of an extracellular domain of leucine-rich repeats, three immunoglobulin-like domains and an intracellular region. The gene to this protein is located on human chromosome 12q13.2. LRIG3 mRNA is predominantly expressed in thyroid, skin and stomach, and is least expressed in heart and blood.

Immunogen

Leucine-rich repeats and immunoglobulin-like domains protein 3 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

LRIG3 (leucine-rich repeats and immunoglobulin-like domains 3) is a cytokine. Its function is not yet fully known. It is either influenced by or involved in influencing the pattern of tumor specific markers in precancerous and cancerous cells. Its expression is also linked with the events in cervical intraepithelial neoplasia (CIN), as well as with the use of hormonal contraceptives. LRIG3 gene lies within the region which is up-regulated in certain types of glioblastomas. Hence, this gene is linked with the pathophysiology and prognosis of glioblastoma. Studies show that over-expression of this gene leads to induction of apoptosis and inhibition of proliferation and invasion in bladder cancer cells. LRIG3 regulates the expression of Netrin1 in the epithelium of otic vesicle. This way, it mediates the formation of semicircular canal.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72023

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Qi et al.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban, 33(1), 111-116 (2013-02-09)
The purpose of this study was to investigate the impact of leucine-rich repeats and immunoglobulin-like domains 3 (LRIG3) on the biological features of bladder cancer cell lines. The plasmids of over-expressed LRIG3 and the blank plasmid serving as control were
Xiaoyi Yuan et al.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban, 31(2), 220-225 (2011-04-21)
This study examined the effect of silencing LRIG3 expression on the proliferation and apoptosis of bladder cancer T24 cells and explored the role of LRIG3 in the tumorigenesis of bladder cancer. Bladder cancer T24 cells were routinely cultured and pSilencer
Victoria E Abraira et al.
PloS one, 5(2), e8981-e8981 (2010-02-04)
The Lrig genes encode a family of transmembrane proteins that have been implicated in tumorigenesis, psoriasis, neural crest development, and complex tissue morphogenesis. Whether these diverse phenotypes reflect a single underlying cellular mechanism is not known. However, Lrig proteins contain
A K Lindström et al.
European journal of histochemistry : EJH, 58(2), 2227-2227 (2014-07-08)
The novel biomarker LRIG3 is a member of the LRIG family (LRIG1-3). While LRIG1 has been associated with favorable prognosis and LRIG2 with poor prognosis in invasive cervical cancer, little is known about the role of LRIG3. The aim of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service