Skip to Content
Merck
All Photos(7)

Key Documents

HPA010083

Sigma-Aldrich

Anti-TIMM17A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-TIM17, Anti-TIM17A, Anti-translocase of inner mitochondrial membrane 17 homolog A (yeast)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on30 April 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Estimated to ship on30 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

TRFASAQFPNGPQFAEDPSQLPSTQLPSSPFGDYRQYQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TIMM17A(10440)

General description

TIMM17A (translocase of inner mitochondrial membrane 17 homolog A) is a member of the TIM23 presequence translocase of the inner mitochondrial membrane complexes. It is highly conserved across eukaryotes. It is one of the two TIM17 genes, and is expressed in adult and fetal tissues, where it forms complexes with TIM23 protein.

Immunogen

translocase of inner mitochondrial membrane 17 homolog A (yeast) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TIMM17A (translocase of inner mitochondrial membrane 17 homolog A) is a part of the inner-membrane complexes, which participate in the membrane insertion of mitochondrial proteins. This protein connects TIM23 with PAM complex, and is responsible for the dynamics of TIM23 complex. Inactivation of this protein leads to the collapse of TIM23 channel into a single pore. Thus, it is essential for maintaining pore structure and voltage gating of TIM23 channels. It prevents mitochondrial (mt)DNA instability and hence, prevents mtDNA loss. In human breast cancers, its expression is linked with unfavorable parameters including, tumor grade, NPI (Nottingham prognostic index), TNM stage and nodal positivity. Thus, in breast cancer, its expression is linked with poor prognosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71954

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michelina Iacovino et al.
Human molecular genetics, 18(1), 65-74 (2008-10-02)
Maintenance of an intact mitochondrial genome is essential for oxidative phosphorylation in all eukaryotes. Depletion of mitochondrial genome copy number can have severe pathological consequences due to loss of respiratory capacity. In Saccharomyces cerevisiae, several bifunctional metabolic enzymes have been
Mohamed Salhab et al.
Breast cancer (Tokyo, Japan), 19(2), 153-160 (2010-10-26)
Mitochondrial dysfunction can be associated with genomic instability and has been implicated in the pathogenesis of several human malignancies, including breast cancer (BC). The mitochondrial protein, translocase of inner mitochondrial membrane 17 homolog A (TIMM17A) contributes to a pre-protein import
Xiaoen Xu et al.
Proteomics, 10(7), 1374-1390 (2010-03-04)
The proteins involved in breast cancer initiation and progression are still largely elusive. To gain insights into these processes, we conducted quantitative proteomic analyses with 21T series of breast cell lines, which include a normal, primary tumor and a metastatic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service