Skip to Content
Merck
All Photos(3)

Key Documents

HPA009687

Sigma-Aldrich

Anti-PIP5K1B antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PIP5KIbeta, Anti-Phosphatidylinositol-4-phosphate 5-kinase type I beta, Anti-Phosphatidylinositol-4-phosphate 5-kinase type-1 beta, Anti-Protein STM-7, Anti-PtdIns(4)P-5- kinase beta, Anti-Type I phosphatidylinositol-4-phosphate 5- kinase beta

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on03 May 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Estimated to ship on03 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

YFDTETYNALMKTLQRDCRVLESFKIMDYSLLLGIHFLDHSLKEKEEETPQNVPDAKRTGMQKVLYSTAMESIQGPGKSGDGIITENPDTMGGIPAKSH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PIP5K1B(8395)

General description

PIP5K1B (phosphatidylinositol-4-phosphate 5-kinase, type I β) is one of the three PIPKI isoforms and is equivalent to mouse PIP5K1α. Each isoform has distinct fibroblast localization. This protein resides in the uropod of polarized dHL60 cells, which requires its C-terminal.

Immunogen

Phosphatidylinositol-4-phosphate 5-kinase type-1 beta recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PIP5K1B (phosphatidylinositol-4-phosphate 5-kinase, type I β) is essential for the control of leukocyte motility by arranging signaling pathways at the cell posterior. Inactivation of this protein results in aberrant cell polarity and directionality during chemotaxis. It functions as a scaffold for interaction among multiple proteins at the uropod. This protein mainly converts phosphatidylinositol 4-phosphate to phosphatidylinositol 4,5-bisphosphate (PI4,5P2). This protein controls the disassembly of focal adhesion, and might be responsible for the endocytosis of integrin at cell adhesion sites, which eventually impacts cell migration.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72473

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Ting Chao et al.
Molecular and cellular biology, 30(18), 4463-4479 (2010-07-14)
Cell migration requires the regulated disassembly of focal adhesions, but the underlying mechanisms remain poorly defined. We have previously shown that focal adhesion disassembly requires the dynamin 2- and clathrin-dependent endocytosis of ligand-activated beta1 integrins. Here, we identify type I
Rosa Ana Lacalle et al.
The Journal of cell biology, 179(7), 1539-1553 (2007-12-26)
Directional cell movement in response to external chemical gradients requires establishment of front-rear asymmetry, which distinguishes an up-gradient protrusive leading edge, where Rac-induced F-actin polymerization takes place, and a down-gradient retractile tail (uropod in leukocytes), where RhoA-mediated actomyosin contraction occurs.
Danila Di Meo et al.
Life science alliance, 7(5) (2024-03-05)
During their differentiation, neurons establish a highly polarized morphology by forming axons and dendrites. Cortical and hippocampal neurons initially extend several short neurites that all have the potential to become an axon. One of these neurites is then selected as
Rosa Ana Lacalle et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(6), 2371-2385 (2015-02-26)
Type I phosphatidylinositol 4-phosphate 5-kinases (PIP5KIs; α, β, and γ) are a family of isoenzymes that produce phosphatidylinositol 4,5-bisphosphate [PI(4,5)P2] using phosphatidylinositol 4-phosphate as substrate. Their structural homology with the class II lipid kinases [type II phosphatidylinositol 5-phosphate 4-kinase (PIP4KII)]

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service