Skip to Content
Merck
All Photos(3)

Documents

HPA008888

Sigma-Aldrich

Anti-HSF1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HSF 1 antibody produced in rabbit, Anti-HSTF 1 antibody produced in rabbit, Anti-Heat shock factor protein 1 antibody produced in rabbit, Anti-Heat shock transcription factor 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SNVPAFLTKLWTLVSDPDTDALICWSPSGNSFHVFDQGQFAKEVLPKYFKHNNMASFVRQLNMYGFRKVVHIEQGGLVKPERDDTEFQHPCFLRGQEQLLENIKRKVTSVSTLKSEDIKIRQDSVTKLLTDVQLMKGKQECMDSKLLA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSF1(3297)

General description

HSF1 (heat shock transcription factor 1) is a transcriptional protein, which plays an essential role in the heat shock process. It is a multi-domain protein, which is composed of a helix winged-loop helix DNA binding domain (DBD) in its N-terminal, three leucine zipper domains (LZ1-3), a core regulatory domain, another leucine zipper domain (LZ4), and a C-terminal transcription activation domain.

Immunogen

Heat shock factor protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

HSF1 (heat shock transcription factor 1) is capable of controlling the transcription of other genes. It mediates the expression of chaperone proteins, and increases cell survival under thermal stress. It regulates protein homeostasis, thus, determining ageing and longevity. It also functions as a transcription regulator of dystrophin protein, Dp71. This protein is over-expressed in hepatocellular carcinoma (HCC), and thus, has potential as a prognostic marker as well as a therapeutic target in the same. This protein is activated in CAFs (cancer associated fibroblasts), where it promotes maliganancy. Thus, high stromal expression of HSF1 protein in early-stage lung and breast is linked with poor patient prognosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71034

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruth Scherz-Shouval et al.
Cell, 158(3), 564-578 (2014-08-02)
Stromal cells within the tumor microenvironment are essential for tumor progression and metastasis. Surprisingly little is known about the factors that drive the transcriptional reprogramming of stromal cells within tumors. We report that the transcriptional regulator heat shock factor 1 (HSF1)
Jin Tan et al.
Cell stress & chaperones, 20(2), 371-379 (2014-11-29)
Heat shock factor 1 (HSF1) is one of the most important transcriptional molecules in the heat shock process; however, HSF1 can also regulate the expression of other proteins. Dystrophin Dp71 is one of the most widely expressed isoforms of the
Shulian Li et al.
Molecular medicine reports, 10(5), 2313-2321 (2014-09-10)
Heat shock factor 1 (HSF1) is associated with tissue‑specific tumorigenesis in a number of mouse models, and has been used a as prognostic marker of cancer types, including breast and prostatic cancer. However, its role in human hepatocellular carcinoma (HCC) is
Felicia K Ooi et al.
Science signaling, 10(501) (2017-10-19)
Learning, a process by which animals modify their behavior as a result of experience, enables organisms to synthesize information from their surroundings to acquire resources and avoid danger. We showed that a previous encounter with only the odor of pathogenic
Alex M Jaeger et al.
The Journal of biological chemistry, 289(44), 30459-30469 (2014-09-11)
The heat shock transcription factor 1 (HSF1) activates expression of a variety of genes involved in cell survival, including protein chaperones, the protein degradation machinery, anti-apoptotic proteins, and transcription factors. Although HSF1 activation has been linked to amelioration of neurodegenerative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service