Skip to Content
Merck
All Photos(5)

Key Documents

HPA008716

Sigma-Aldrich

Anti-RNF14 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Androgen receptor-associated protein 54, Anti-E3 ubiquitin-protein ligase RNF14, Anti-HFB30, Anti-RING finger protein 14, Anti-Triad2 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on28 May 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$1,040.00


Estimated to ship on28 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

GCTMGICSSCNFAFCTLCRLTYHGVSPCKVTAEKLMDLRNEYLQADEANKRLLDQRYGKRVIQKALEEMESKEWLEKNSKSCPCCGTPIEKLDGCNKMTCTGCMQYFCWICMGSLSRANPYKHFNDPGSPCFNRLFYAVDVDDDIWEDEV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF14(9604)

General description

RNF14 (ring finger protein 14) is a ligand-dependent androgen receptor (AR)-associated protein and is also called ARA54. It has a molecular weight of 54kDa and is composed of 474 amino acids. Its middle portion contains RING finger or zinc finger motif, which is rich in cysteine. Downstream to the RING finger motif, it contains another cysteine-rich region similar to B-box. This protein might be the representative of a new subgroup of the B-box RING finger protein family. Its mRNA has the highest expression in testis, followed by thymus, spleen, colon, prostate, uterus, small intestine, and leukocytes of blood. This gene is localized to human chromosome 5q23.3-q31.1.

Immunogen

E3 ubiquitin-protein ligase RNF14 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RNF14 (ring finger protein 14), along with ARA70 or SRC-1 (steroid receptor coactivator), co-activates AR (androgen receptor), and acts as the preferred AR activator in prostate cancer DU145 cells. Thus, it might have an important role in AR-signaling pathway in prostate. It acts as an activator of cyclin D1, which promotes the progression of cell cycle and cell proliferation in human tumor cells. This protein acts as a positive regulator of Wnt signaling in cancer cells, and is essential for colon cancer cell survival. It also acts as a regulator of T-cell factor/lymphoid enhancer factor (TCF/LEF), which in turn control transcription through the recruitment of β-catenin. Spermatogenesis and testicular development in humans is regulated by the localization patter on RNF14 protein, and this protein is down-regulated in non-obstructive azoospermia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86694

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Ueki et al.
Biochimica et biophysica acta, 1445(2), 232-236 (1999-05-13)
A human cDNA, HFB30, encoding a novel protein that contains a RING finger (C3HC4-type zinc finger) motif was isolated. This cDNA clone consists of 3056 nucleotides and encodes an open reading frame of a 474 amino acid protein. From RT-PCR
Kuo-Chung Lan et al.
Fertility and sterility, 89(5 Suppl), 1397-1405 (2007-10-09)
To elucidate the physiologic and pathologic roles of androgen receptor (AR) and co-regulators in human testes with obstructive azoospermia or nonobstructive azoospermia. Prospective laboratory and clinical study. Infertility clinic at Chang Gung Memorial Hospital. Twenty-seven men with obstructive azoospermia and
H Y Kang et al.
The Journal of biological chemistry, 274(13), 8570-8576 (1999-03-20)
Androgen receptor (AR) is a member of the steroid receptor superfamily that may require coactivators for proper or maximal transactivation. Using a yeast two-hybrid screening followed by mammalian cell analyses, we identified a novel ligand-dependent AR-associated protein, ARA54, which consists
Beibei Wu et al.
EMBO reports, 14(4), 347-355 (2013-03-02)
T-cell factor/lymphoid enhancer factor (TCF/LEF) proteins regulate transcription by recruiting β-catenin and its associated co-regulators. Whether TCF/LEFs also recruit more factors through independent, direct interactions is not well understood. Here we discover Ring Finger Protein 14 (RNF14) as a new
Hirotoshi Kikuchi et al.
Carcinogenesis, 28(8), 1752-1758 (2007-05-19)
Cyclin D1 is one of the major enhancers of cell cycle progression and its expression is regulated in several growth stimulatory signaling pathways. ARA54 is an androgen receptor (AR) co-activator that enhances AR-dependent transcriptional activation. Although expression of ARA54 mRNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service