Skip to Content
Merck
All Photos(7)

Key Documents

HPA008440

Sigma-Aldrich

Anti-PANK2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Pantothenate kinase 2, mitochondrial precursor antibody produced in rabbit, Anti-Pantothenic acid kinase 2 antibody produced in rabbit, Anti-hPANK2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on31 May 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Estimated to ship on31 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

SLHTVFCATGGGAYKFEQDFLTIGDLQLCKLDELDCLIKGILYIDSVGFNGRSQCYYFENPADSEKCQKLPFDLKNPYPLLLVNIGSGVSILAVYSKDNYKRVTGT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PANK2(80025)

General description

Pantothenate kinase 2 (PANK2) is localized to the mitochondria. The gene encoding the enzyme is present on chromosome 20.

Immunogen

Pantothenate kinase 2, mitochondrial precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Pantothenate kinase 2 (PANK2) plays an important role in energy metabolism and in the synthesis of coenzyme A (CoA) from pantothenate. It phosphorylates pantothenate to 4′-phosphopantothenate, which then exits the mitochondria because the enzymes catalyzing further reactions in the CoA biosynthetic pathway are cytosolic. It is finally converted to CoASH. Mutations in this gene are associated with PanK-associated neurodegeneration (PKAN), characterized by pathological accumulation of iron in the basal ganglia, dystonia, dysarthria, intellectual impairment, and gait disturbance.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71386

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yien-Ming Kuo et al.
Human molecular genetics, 14(1), 49-57 (2004-11-05)
Pantothenate kinase-associated neurodegeneration (PKAN, formerly known as Hallervorden-Spatz syndrome) is a rare but devastating neurodegenerative disorder, resulting from an inherited defect in coenzyme A biosynthesis. As pathology in the human condition is limited to the central nervous system, specifically the
Roberta Leonardi et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(5), 1494-1499 (2007-01-24)
The human isoform 2 of pantothenate kinase (PanK2) is localized to the mitochondria, and mutations in this protein are associated with a progressive neurodegenerative disorder. PanK2 inhibition by acetyl-CoA is so stringent (IC50 < 1 microM) that it is unclear
Neil Gordon
European journal of paediatric neurology : EJPN : official journal of the European Paediatric Neurology Society, 6(5), 243-247 (2002-10-11)
The arguments over the nomenclature of the syndrome are reviewed. Ethical considerations favour replacing the present eponyms with the title of panthothenate kinase-associated neurodegeneration (PKAN), now that more is known about the cause of the condition. The symptoms and signs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service