Skip to Content
Merck
All Photos(3)

Key Documents

HPA008266

Sigma-Aldrich

Anti-PFKFB3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3, Anti-6PF-2-K/Fru-2,6-P2ASE brain/placenta-type isozyme, Anti-Renal carcinoma antigen NY-REN-56, Anti-iPFK-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Available to ship on14 April 2025Details



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$1,040.00


Available to ship on14 April 2025Details


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 1:100-1:250
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

EEAMKVRKQCALAALRDVKSYLAKEGGQIAVFDATNTTRERRHMILHFAKENDFKAFFIESVCDDPTVVASNIMEVKISSPDYKDCNSAEAMDDFMKRISCYEASYQPLDPDKCDRDLSLIKVIDVGRRFLVNRVQD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PFKFB3(5209)

General description

The gene encoding the protein fructose-2, 6-bisphosphatase 3 (PFKFB3) is localized on human chromosome 10p15-p14. It is expressed in activated lymphoid cells and in different tumor cell lines.

Immunogen

6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Fructose-2, 6-bisphosphatase 3 (PFKFB3) is a bifunctional enzyme which controls amount of fructose-2,6-bisphosphate, an activator of phosphofructokinase in glycolysis. PFKFB3 controls the glycolytic flux to lactate and the non-oxidative pentose shunt.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71520

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hidenori Bando et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 11(16), 5784-5792 (2005-08-24)
Fructose 2,6-bisphosphate (F2,6BP) is a potent activator of phosphofructokinase, which is a rate-limiting enzyme of glycolysis. The concentration of F2,6BP depends on the activity of the bifunctional enzyme, 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase (PFK-2/FBPase). Four genes encoding PFK-2/FBPase have been identified and termed PFKFB1
Jason Chesney
Current opinion in clinical nutrition and metabolic care, 9(5), 535-539 (2006-08-17)
Neoplastic cells metabolize abundant glucose relative to normal cells in order to satisfy the increased energetic and anabolic needs of the transformed state. This review will summarize the requirement of 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatases for the regulation of glycolysis in cancer cells and
Ulrich Mahlknecht et al.
International journal of oncology, 23(4), 883-891 (2003-09-10)
PFKFB (6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase) is a bifunctional enzyme that regulates the steady-state concentration of fructose-2,6-bisphosphate, which is a potent activator of the key regulatory enzyme of glycolysis, phosphofructokinase. PFKFB3 (iPFK2) is one of four tissue-specific PFKFB isozymes that have been identified to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service