Skip to Content
Merck
All Photos(9)

Key Documents

HPA008150

Sigma-Aldrich

Anti-TMPO antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Lamina-associated polypeptide 2, isoforms β/γ antibody produced in rabbit, Anti-TP β/γ antibody produced in rabbit, Anti-TPRP isoforms β/γ antibody produced in rabbit, Anti-Thymopoietin, isoforms β/γ antibody produced in rabbit, Anti-Thymopoietin-related peptide isoforms β/γ antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Estimated to ship on30 May 2025



Select a Size

Change View
100 μL
$952.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$952.00


Estimated to ship on30 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

enhanced validation

orthogonal RNAseq
RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VNPGPIVGTTRKLYEKKLLKLREQGTESRSSTPLPTISSSAENTRQNGSNDSDRYSDNEEDSKIELKLEKREPLKGRAKTPVTLKQRRVEHNQSYSQAGITETEWTSGSSKGGPLQALTRESTRGSRRTPRKRVETSE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TMPO(7112)

General description

TMPO (thymopoietin) gene is mapped to human chromosome 12q22. Alternate splicing of the mRNA results in isoforms α (75kDa), β (51kDa), and γ (39kDa), which are related nuclear proteins. The gene consists of eight exons spanning a length of ∼35kb. Exon 4 is spliced into α mRNA, which encodes a putative basic nuclear localization motif. Exon 8 encodes a hydrophobic putative membrane-spanning domain that targets β and γ isoforms to the nuclear membrane

Immunogen

Lamina-associated polypeptide 2, isoforms β/γ recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TMPO (thymopoietin) gene is also called as LAP2 (Lamina-associated polypeptide 2) and functions in the assembly of the nuclear lamina. It is present in the nucleus and facilitates the maintenance of structural organization of the nuclear envelope. It serves as an anchor for the attachment of lamin filaments to the inner nuclear membrane.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71860

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C A Harris et al.
Genomics, 28(2), 198-205 (1995-07-20)
Thymopoietins (TMPOs, previously abbreviated TPs) alpha (75 kDa), beta (51 kDa), and gamma (39 kDa) are related nuclear proteins expressed in many or all tissues. TMPO alpha is present diffusely throughout the nucleus, while TMPOs beta and gamma are localized
Razan A Elkhatib et al.
Reproduction (Cambridge, England), 154(4), 387-401 (2017-07-08)
During spermiogenesis the spermatid nucleus is elongated, and dramatically reduced in size with protamines replacing histones to produce a highly compacted chromatin. After fertilisation, this process is reversed in the oocyte to form the male pronucleus. Emerging evidence, including the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service