Skip to Content
Merck
All Photos(5)

Key Documents

HPA007656

Sigma-Aldrich

Anti-CAMKV antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CaM kinase-like vesicle-associated protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

LLSGNPPFYEEVEEDDYENHDKNLFRKILAGDYEFDSPYWDDISQAAKDLVTRLMEVEQDQRITAEEAISHEWISGNAASDKNIKDGVCAQIEKNFARAKW

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAMKV(79012)

Immunogen

CaM kinase-like vesicle-associated protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CAMKV (CaM kinase-like vesicle-associated) gene encodes a member of the CaMK (Ca2+/calmodulin-dependent protein kinases) superfamily that mediate stimulus–secretion coupling and excitation–contraction coupling in various cell types. The protein is a monomer of 41kDa that is 504 amino acids long and is found to be associated with perikaryal membranes and neurites. This calmodulin binding protein is also associated with vesicles in both axons and dendrites. It is found to be associated with membranes and vesicles present in neurites of subsets of noncerebellar neurons. The N-terminal domain of this protein exhibits high similarity to the catalytic and calmodulin-binding domains of CaMKII, another member of the same family. The C-terminus contains an eight-residue motif DR/GSA/VTPAT that may be related to PEST sequences that confer half-life to proteins in which they are present. It contains a 19-residue hydrophobic region that may span a membrane and a 32-residue uncharged region. This protein resembles other members of the family and may function as a calmodulin-binding protein kinase.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71444

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Yoshida et al.
Biochimica et biophysica acta, 1497(1), 155-167 (2000-06-06)
Ca(2+)/calmodulin-dependent protein kinases (CaMKs) are important intracellular mediators in the mediation of stimulus-secretion coupling and excitation-contraction coupling in a wide variety of cell types. We attempted to identify and characterize the functional roles of CaMK in mediating pancreatic enzyme secretion.
M Godbout et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 14(1), 1-13 (1994-01-01)
We have characterized cDNA clones of 1G5, an mRNA highly enriched in the mammalian forebrain that encodes a 504-residue protein found in association with perikaryal membranes and neurites. The protein, which accumulates predominantly postnatally, is associated with vesicles in both

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service