Skip to Content
Merck
All Photos(8)

Key Documents

HPA007571

Sigma-Aldrich

Anti-GLUL antibody produced in rabbit

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GS antibody produced in rabbit, Anti-Glutamate--ammonia ligase antibody produced in rabbit, Anti-Glutamine synthetase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Estimated to ship on30 May 2025



Select a Size

Change View
100 μL
$952.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$952.00


Estimated to ship on30 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

RVCEDFGVIATFDPKPIPGNWNGAGCHTNFSTKAMREENGLKYIEEAIEKLSKRHQYHIRAYDPKGGLDNARRLTGFHETSNINDFSAGVANRSASIRIPRTVGQEKKGYFEDRRPSANCDPFSVTEALIRTCLLNETG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GLUL(2752)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

GLUL (glutamate-ammonia ligase) gene encodes an ATP-dependent enzyme that catalyzes the synthesis of glutamine from glutamate and ammonia (nitrogen source). It is a member of the glutamine synthetase family.

Immunogen

Glutamine synthetase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GLUL (glutamate-ammonia ligase) is an essential enzyme in glutamine neogenesis. It functions in detoxification of ammonia and glutamate, acid-base homeostasis, cell signaling, and cell proliferation. It has a role in proliferation of fetal skin fibroblasts. Defects in this gene is associated with congenital glutamine deficiency. Overexpression of this gene has been observed in some primary liver cancer samples.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70154

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruri Tsuneishi et al.
Scientific reports, 11(1), 11381-11381 (2021-06-02)
Ammonia has a cytotoxic effect and can therefore be used as a selection agent for enrichment of zone I hepatocytes. However, it has not yet been determined whether ammonia-treated hepatocyte-like cells are able to proliferate in vitro. In this study
Jong-Ho Choi et al.
Journal of cellular biochemistry, 114(2), 303-313 (2012-08-30)
In our previous study, we screened and isolated genes that were up-regulated after partial pancreatectomy using transcriptomic analysis and glutamate-ammonia ligase (GLUL) was selected for further study based on its effect on differentiation and proliferation. In the immunohistochemical analysis, GLUL
Ying-Hui Ko et al.
Cancer biology & therapy, 12(12), 1085-1097 (2012-01-13)
Glutamine metabolism is crucial for cancer cell growth via the generation of intermediate molecules in the tricarboxylic acid (TCA) cycle, antioxidants and ammonia. The goal of the current study was to evaluate the effects of glutamine on metabolism in the
Kirsten E Fleming et al.
Liver international : official journal of the International Association for the Study of the Liver, 33(4), 525-534 (2013-02-01)
In normal human liver, glutamine synthetase (GS) is expressed in a rim of hepatocytes surrounding hepatic veins. GS expression is decreased in cirrhosis and increased in chronic hepatitis, focal nodular hyperplasia, peritumoural hyperplasia and some hepatocellular neoplasms. For the non-neoplastic
Johannes Häberle et al.
The New England journal of medicine, 353(18), 1926-1933 (2005-11-04)
Glutamine synthetase plays a major role in ammonia detoxification, interorgan nitrogen flux, acid-base homeostasis, and cell signaling. We report on two unrelated newborns who had congenital human glutamine synthetase deficiency with severe brain malformations resulting in multiorgan failure and neonatal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service