Skip to Content
Merck
All Photos(1)

Key Documents

HPA007386

Sigma-Aldrich

Anti-PRKG2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CGK 2 antibody produced in rabbit, Anti-Type II cGMP-dependent protein kinase antibody produced in rabbit, Anti-cGKII antibody produced in rabbit, Anti-cGMP-dependent protein kinase 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

REYHLKELREQLSKQTVAIAELTEELQNKCIQLNKLQDVVHMQGGSPLQASPDKVPLEVHRKTSGLVSLHSRRGAKAGVSAEPTTRTYDLNKPPEFSFEKARVRKDSSEKKLITDALNKNQFLKRLDPQQIKDMVE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRKG2(5593)

General description

PRKG2 is also known as cGK II (Type II cGMP-dependent protein kinase) that contains an N-terminal regulatory domain, two cGMP-binding domains and a C-terminal catalytic domain. The N-terminal domain contains a dimerization and an autoinhibitory region. It is found to be bound to the membrane by a myristoyl moiety. It is mainly expressed in intestine, brain, and kidney. The gene is localized to human chromosome 4 and contains 19 exons spanning a length of 125kb.

Immunogen

cGMP-dependent protein kinase 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PRKG2 (protein kinase, cGMP-dependent, type II) gene encodes a serine/threonine protein kinase that functions in the regulation of intestinal fluid balance in human. It regulates the electrolyte and water secretion by epithelial tissues when stimulated by luminocrinic hormones guanylin and uroguanylin. It functions in secretory diarrhea triggered by heat-stable enterotoxins. It also plays an important role in several processes such as endochondral ossification by C-type natriuretic peptide, renin secretion by the kidney, aldosterone secretion by the adrenal and adjustment of the circadian rhythm.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86393

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirja Koch et al.
International journal of molecular sciences, 22(1) (2020-12-31)
Mutations in the CNGA3 gene, which encodes the A subunit of the cyclic guanosine monophosphate (cGMP)-gated cation channel in cone photoreceptor outer segments, cause total colour blindness, also referred to as achromatopsia. Cones lacking this channel protein are non-functional, accumulate
Arie B Vaandrager et al.
Frontiers in bioscience : a journal and virtual library, 10, 2150-2164 (2005-06-23)
Type II cGMP-dependent protein kinase (cGK II) is the protein product of one of two genes coding for cGKs in mammalian genomes. cGK II has a domain structure similar to cGK I (alpha or beta) consisting of an N-terminal regulatory
O Witczak et al.
Biochemical and biophysical research communications, 245(1), 113-119 (1998-05-16)
The type II cGMP-dependent protein kinase (cGK) plays a pivotal role in the regulation of intestinal fluid balance in man. Furthermore, mice carrying a null mutation for the gene encoding the type II cGK develop as dwarfs indicating that this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service